Oxoid™ Perfringens TSC Supplement
Oxoid™ Perfringens TSC Supplement
Thermo Scientific™
Oxoid™ Perfringens TSC Supplement
Oxoid Perfringens TSC Supplement (D-cycloserine 200mg/vial) is used for the recovery of Clostridium perfringens.
Have Questions?
Catalog number SR0088E
Price (USD)/ Each
-
Request A Quote
Quantity:
10/Pk.
Oxoid Perfringens TSC Supplement (D-cycloserine 200mg/vial) is used for the recovery of Clostridium perfringens.
  • Add to DCLS Agar, Part No. CM0393B, or Perfringens Agar Base, TSC, Part No. CM0587B
  • Each vial supplements 500mL of medium
For Laboratory Use
For Laboratory Use
Specifications
SpeciesPerfringens
DescriptionPerfringens TSC Supplement
FormFreeze-dried
Product TypeMicrobiology Media
Quantity10/Pk.
YieldEach vial for 500mL medium
Unit SizeEach