Industry expertise in process analysis
Water quality is critical to the health of our communities, environment, and operations. Thermo Fisher Scientific has been advising and supporting companies worldwide on process analytics. Key competencies are the process-safe measurement of pH, ORP, conductivity, and oxygen.
In addition to sensors, we also offer innovative process analysers, high-quality holders and fittings, and unique cleaning and calibration systems.
Monitor your water quality with our process analytics solutions in the pharmaceutical, chemical, food, water, and energy industries.
The critical parameters of process-safe liquids analysis in the pharmaceutical industry are pH, oxygen, and conductivity.
Deviations from given measurement values could have an impact on production results. A broad portfolio of sensors, process analysers, holders and fittings, cleaning and calibration systems allows us to offer tailor-made solutions and services.
The pharmaceutical sensors must be sterilisable in place (SIP) and cleanable in place (CIP). We provide a wide range of advanced Memosens sensors from Knick that can be easily connected to the MemoSuite advanced asset management tool. This system helps track changes in the process or measurement and ensures that data cannot be manipulated.
Process safe liquids analysis is critical to the chemical industry, with critical parameters such as oxygen, pH, and conductivity.
Measuring points are often exposed to rough physical and chemical conditions that can rapidly degrade standard connections between sensors, cables, and transmitters. The rugged Memosens digital inductive sensor connections are free of any metallic contacts. This results in a plug-and-play system that is 100% waterproof and resistant to dirt, corrosion, signal interference and is exceptionally resilient to mechanical influences.
IECEx explosion area rated versions of products are available for those zones that require it, including the Portavo range of portable meters.
We have a range of rugged fittings and sensors that monitor critical parameters of pH, oxygen, and conductivity even when the process includes aggressive reactants/intermediates, highly toxic production processes, and abrasive slurries in industries such as manufacturing, refining, and extraction.
Monitoring critical parameters of pH, oxygen, and conductivity span the entire food and beverage production process from pure water used in the manufacturing process to water used in cleaning to wastewater discharged from the plant.
We offer a broad portfolio of sensors, process analysers, holders and fittings, cleaning, and calibration systems from Knick that address FDA requirements within the production line.
High purity water is an essential and integral part of energy production. Similarly, the prevention of corrosion and mineral deposits is critical to reducing wear and downtime. Monitoring pH, oxygen, and conductivity is key to detecting potential issues.
Oxygen is highly corrosive; even minimal concentrations in the ppm range can corrode metal surfaces. Increased conductivity is a reliable indication of contamination from salts, minerals, and other contaminants. Mineral deposits and corrosion reduce efficiency and lead to the premature wear of system components. Also, deposits of lime or corrosion provide a platform for organic material and biofilm.
pH, oxygen, and conductivity monitoring is core to all water quality control processes, from drinking water supply to wastewater treatment.
Online measurement of pH value and oxidation-reduction potential (ORP) enables operators in drinking water treatment plants to assess the quality of the raw water they use and implement measures to protect the system.
Equally important is monitoring pH, DO, and conductivity at multiple points during wastewater treatment. Plants can have harsh environments that can rapidly degrade regular connections between sensors, cables, and transmitters, as well as taking a toll on exposed transmitters or controllers. Our high IP protection rated enclosures, combined with Memosens inductive connection and transmission technology, ensures your investment continues to deliver reliable service year after year.
Portable meters
The first portables with Memosens technology for measuring pH / ORP, conductivity, and oxygen comes from Knick - the only provider of mobile, portable meters for hazardous locations.
Suitable for use in industry, in the lab, for environmental protection, food processing, and water or wastewater treatment, the Portavo range of meters feature a graphical colour display, data logger, USB port, and Li-ion battery.
The ultra-flat yet robust housing is protected against strong jets of water and fits comfortably in hand, while the integrated sensor quiver protects the sensor against drying out.
Industrial transmitters
Industrial transmitters for pH, ORP, conductivity, and oxygen meters. These are available in 2- or 4-wire configuration, explosion-protected.
Bus communication: HART, FOUNDATION Fieldbus, Profibus DP, and PA.
Robust industrial housing with high IP protection for outdoor applications.
The transmitters are available with analog and digital sensors and are easy and intuitive to use operation.
From the DIN rail transmitter to the fully modular system, the product range includes the Protos, Stratos, and MemoRail, families.
Sensor holders and fittings
Holders and fittings from Knick provide safety and convenience when it comes to measuring pH, ORP, dissolved oxygen and conductivity.
The product range extends from immersion, inline and flow-through fittings over retractable fittings up to fully automatic calibration and cleaning systems.
Also included in the portfolio are Knick retractable fittings for fibre optic sensors. Whether PVDF, PEEK or stainless steel –fittings are available in a variety of materials suitable for every process application, and are the first choice for chemicals, pharmaceuticals, paints, oil and sugar.
Industrial sensors
For measuring pH, ORP, conductivity or oxygen in industrial applications, we offer analog as well as digital Memosens-sensors.
They enable low-maintenance and highly precise measurements in the chemical, pharmaceutical and food industries, energy and environment.
Resources
Brochure: Solutions for the pharmaceutical industry
Brochure: Advanced Liquid Analysis
Brochure:Portavo Portable Multiparameter Analyser
Brochure: Sensors
Brochure:MemoView mobile tool
Brochure:StratosPro2-wire multiparameter analysers for pH/ORP, conductivity or oxygen
Brochure:StratosEvo 4-wire multiparameter analyser for pH/ORP, conductivity or oxygen
Brochure:StratosEco
Brochure:StratosMS
Brochure:MemoRail Modbus A1405 N
Brochure:SensoGate retractable fittings
Brochure:Ceramat retractable fittings for extreme conditions
Brochure:ProtosII Modular Premium Transmitter