Sandra Nason
Technical Support Specialist
Environmental and Process Monitoring
Meet our techs: Sandra Nason
Sandra has been working for Thermo Fisher Scientific since 2007 and brings her expertise in electronics, production and testing towards quick and comprehensive solutions for our customers.
Our techs are here to answer your questions, just email us.
If you like this month's Q&A, check out our Talk Tech archive.
Question
The -273°C number is absolute zero and it means that there is an open circuit. First, please review the following Technical Bulletin to find instructions on how to check the cable connections for corrosion. Then use the information in this technical bulletin to reinstall the new probe and/or cable if necessary. Lastly, perform the following steps:
- Detach the ambient temp probe (with radiation shield) from the 10m cable and connect it directly to the back of the 1405 TEOM™, Continuous Ambient Particulate Monitor.
- If you have a solid connection here, the cable needs to be replaced. (Part #: 51-010812)
- If the temperature reading is still -273°C, the sensor should be changed out with a known good one, replacement sensor cable. (Part #: 51-010792)
- If with a known good sensor attached to the back of the unit, the temperature is still 273°C, please contact technical support:
Telephone: 866-282-0430
International Telephone: 508-520-0430
Select menu option 2 for Technical Support
epm.techsupport@thermofisher.com
To order spare parts, call (866) 282-0430, option 1 or fill out the part request form for a quote.
For more information on this or any other particulate ambient instrument questions, comments or concerns please don’t hesitate to contact me directly.
If you have additional questions, call the Technical Support team at (866) 282-0430 and select option 2.
Don't miss out. Subscribe today.
Talk Tech is published here every month. Receive our monthly Talk Tech answers delivered right to your inbox by subscribing using the button below.