">
LABScreen Screening
Thermo Scientific LABScreen Mixed, Multi, and PRA Assays offer options for detecting and monitoring antibodies, providing versatility for screening and testing policies.
LABScreen Single Antigen
Designed to identify HLA class I and class II antibodies that are reactive to one or more dominant epitopes, Thermo Scientific LABScreen Single Antigen provides a comprehensive panel of antigens to evaluate common and well-documented alleles.
LABScreen Single Antigen ExPlex
Building on the trusted Thermo Scientific LABScreen Single Antigen platform, ExPlex offers an expansive HLA antibody panel in a single-well setup.
C1qScreen
The presence of complement-binding (C1q) antibodies can be identified and monitored in sera, providing information for building better antibody profiles.
LABScreen Autoantibody
Thermo Scientific LABScreen Autoantibody Assays detect and monitor a broad range of autoantibody targets in human sera, utilizing single antigen bead technology.
Not all products are CE marked or have 510(k) clearance for sale in the U.S. Availability of products in each country depends on local regulatory marketing authorization status.
">