Bovine pulmonary artery endothelial cells stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and the anti–cdc6 peptide antibody.
Fixed and permeabilized bovine pulmonary artery endothelial cells stained with Alexa Fluor® 350 phalloidin (Cat. no. A22281), an anti–a-tubulin antibody (Cat. no. A11126) and the anti–cdc6 peptide antibody (Cat. no. A21286). The anti–a-tubulin antibody was labeled with the Zenon® Alexa Fluor® 568 Mouse IgG1 Labeling Kit (Cat. no. Z25006) and the anti–cdc6 peptide antibody was labeled with the Zenon® Alexa Fluor® 488 Mouse IgG1 Labeling Kit (Cat. no. Z25002).
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Fixed and permeabilized bovine pulmonary artery endothelial cells stained with Mouse anti-ATP Synthase Subunit IF1 Monoclonal Antibody. Go ›