Prostate carcinoma.
Tissue from a prostate carcinoma has been fixed with formalin, embedded in paraffin, sectioned, and hybridized with a biotinylated antisense RNA probe to gastrin-releasing peptide (GRP) receptor mRNA. Following in situ hybridization, the biotinylated probe was developed for visualization with alkaline phosphatase&endash;mediated techniques using the ELF 97 mRNA In Situ Hybridization Kit (Cat. No. E6604, E6605). Image contributed by Marty Bartholdi, Berlex Biosciences, Berlex Laboratories, Inc.
Related Products
Related Images
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Labeled paint probes hybridized to human metaphase chromosomes Go ›
Transformed mouse fibroblasts (CRE BAG 2 cells). Go ›
Zebrafish brain. ELF 97 Endogenous Phosphatase Detection Kit. Go ›