Bovine pulmonary artery endothelial cells.
Bovine pulmonary artery endothelial cells were labeled with Alexa Fluor® 488 phalloidin (Cat. No. A12379) to stain F-actin and our mouse monoclonal anti–a-tubulin antibody (Cat. No. A11126) in combination with Alexa Fluor® 594 dye–conjugated F(ab')2 fragment of goat anti–mouse IgG antibody (Cat. No. A11020) to stain microtubules. The multiple-exposure image was acquired using bandpass filter sets appropriate for Texas Red® dye and fluorescein.
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›