A mixture of blue fluorescent, red fluorescent and yellow-green fluorescent FluoSpheres® 4 µm sulfate microspheres.
FluoCells prepared slide #6 (F35925) showing a fixed, permeabilized, and labeled muntjac skin fibroblast. Mitochondria were labeled with mouse anti–OxPhos Complex V inhibitor protein antibody and visualized using orange-fluorescent Alexa Fluor 555 goat anti–mouse IgG antibody (A21422). F-actin was labeled with green-fluorescent Alexa Fluor 488 phalloidin (A12379), and the nucleus was stained with TO-PRO-3 iodide (T3605, pseudocolored magenta).