Immunocytochemistry analysis using Pax 3 (HUP2), Rabbit mAb (16H22L10), ABfinity™ Recombinant Monoclonal Antibody.
Immunocytochemistry analysis of U2OS cells stained with Pax 3 (HUP2), Rabbit mAb (16H22L10), ABfinity™ Recombinant Monoclonal Antibody (Cat. No. 701147), using (A) Alexa Fluor® 488 Goat Anti-Rabbit was used as secondary (green). (B) DAPI was used to stain the nucleus (blue) and (C) Alexa Fluor® 594 phalloidin was used to stain actin (red). (D) Composite image of cells showing nuclear localization of Pax 3.
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
Increase in accuracy and precision using the Qubit® Go ›
Performance of the Qubit® dsDNA HS Assay Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›
Immunocytochemistry analysis using Nucleostemin ABfinity™ Recombinant Rabbit Oligoclonal Antibody. Go ›
Immunocytochemistry analysis using Connexin 40 (C-term) ABfinity™ Recombinant Rabbit Oligoclonal Antibody. Go ›