Neonatal human dermal fibroblasts fixed and permeabilized using the Image-iT® Fixation/Permeabilization Kit.
Human dermal fibroblasts, neonatal (HDFn) (C-004-5C) were fixed and permeabilized using the Image-iT® Fixation/Permeabilization Kit (R37602). Golgi staining was done using an anti-Golgin-97 primary antibody (A21270) and a goat anti-mouse Alexa Fluor® 488 secondary antibody (A11029). Actin was stained using Alexa Fluor® 594 phalloidin (A12381) and nuclei were stained using NucBlue™ Live Cell Stain (R37605). Slides were mounted using ProLong® Gold antifade kit (P7481) and images were acquired on the FLoid™ Cell Imaging Station (4471136).
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Phagocytosis in MMM Cells using pHrodo™ Red E. coli BioParticles® Conjugate® (Catalog # P35361) Go ›
Muntjac skin fibroblast labeled with rhodamine phalloidin. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
Increase in accuracy and precision using the Qubit® Go ›
Performance of the Qubit® dsDNA HS Assay Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
Nuclear, cytoskeletal, and mitochondrial staining of Muntjac fibroblasts. Go ›
Human iPSC staining using mouse anti beta-tubulin monoclonal antibody. Go ›
Human iPSC staining. Go ›