Immunohistochemistry using GFAP Monoclonal Antibody, Mouse, Alexa Fluor® 594 Conjugate
Immunohistochemistry using NMDA Receptor Subunit 2A Polyclonal Antibody, Rabbit: Rat brain cryosections labeled with anti-NMDA receptor, subunit 2A (rat), rabbit IgG fraction (Cat. no.A6473) and detected using Alexa Fluor® 488 goat anti–rabbit IgG antibody (Cat. no. A11008). The tissue was also labeled with Alexa Fluor® 594 anti–glial fibrillary acidic protein antibody (Cat. no.A21295) and counterstained with TOTO®-3 iodide (Cat. no.T3604), which was pseudocolored light blue in this image.
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Schistosoma mansoni parasite. Go ›
The neuromuscular junction of mouse skeletal muscle. Go ›