Immunofluorescent staining using mouse anti-golgin-97 monoclonal antibody
HeLa cells labeled with anti–golgin-97 antibody and detected using Alexa Fluor® 488 Goat Anti–Mouse IgG antibody. Fixed, permeabilized HeLa cells were labeled with anti–golgin-97 monoclonal antibody (A21270) and detected using Alexa Fluor® 488 Goat Anti–Mouse IgG antibody (A11001). The cells were counterstained with DAPI (D1306, D3571, D21490). (Cat. No. A21270).
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›