Amino Acids |
---|
Glycine | 75.0 | 1300.0 | 17.333334 |
L-Alanine | 89.0 | 450.0 | 5.05618 |
L-Arginine hydrochloride | 211.0 | 1400.0 | 6.6350713 |
L-Asparagine | 132.0 | 700.0 | 5.3030305 |
L-Aspartic acid | 133.0 | 700.0 | 5.263158 |
L-Cystine 2HCl | 240.0 | 57.36 | 0.23900001 |
L-Glutamic Acid | 147.0 | 1200.0 | 8.163265 |
L-Glutamine | 146.0 | 1200.0 | 8.219178 |
L-Histidine | 155.0 | 5000.0 | 32.258064 |
L-Isoleucine | 131.0 | 100.0 | 0.7633588 |
L-Leucine | 131.0 | 150.0 | 1.1450381 |
L-Lysine hydrochloride | 183.0 | 1250.0 | 6.830601 |
L-Methionine | 149.0 | 100.0 | 0.67114097 |
L-Phenylalanine | 165.0 | 300.0 | 1.8181819 |
L-Proline | 115.0 | 700.0 | 6.0869565 |
L-Serine | 105.0 | 1100.0 | 10.476191 |
L-Threonine | 119.0 | 350.0 | 2.9411764 |
L-Tryptophan | 204.0 | 200.0 | 0.98039216 |
L-Tyrosine disodium salt | 225.0 | 100.0 | 0.44444445 |
L-Valine | 117.0 | 200.0 | 1.7094017 |
beta-Alanine | 89.0 | 400.0 | 4.494382 |
Vitamins |
---|
Biotin | 244.0 | 0.02 | 8.196721E-5 |
Choline chloride | 140.0 | 0.4 | 0.0028571428 |
D-Calcium pantothenate | 477.0 | 0.04 | 8.385744E-5 |
Folic Acid | 441.0 | 0.04 | 9.0702946E-5 |
Nicotinic acid (Niacin) | 123.0 | 0.04 | 3.2520323E-4 |
Para-Aminobenzoic Acid | 137.0 | 0.04 | 2.919708E-4 |
Pyridoxine hydrochloride | 206.0 | 0.04 | 1.9417475E-4 |
Riboflavin | 376.0 | 0.04 | 1.0638298E-4 |
Thiamine hydrochloride | 337.0 | 0.04 | 1.1869436E-4 |
i-Inositol | 180.0 | 0.04 | 2.2222221E-4 |
Inorganic Salts |
---|
Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 1000.0 | 9.009009 |
Magnesium Chloride (anhydrous) | 95.0 | 2138.64 | 22.512 |
Magnesium Sulfate (MgSO4) (anhyd.) | 120.0 | 2715.34 | 22.627834 |
Potassium Chloride (KCl) | 75.0 | 5600.0 | 74.666664 |
Sodium Bicarbonate (NaHCO3) | 84.0 | 700.0 | 8.333333 |
Sodium Phosphate monobasic (NaH2PO4-H2O) | 138.0 | 2026.0 | 14.681159 |
Other Components |
---|
Alpha-Ketoglutaric acid | 146.0 | 740.0 | 5.0684934 |
D-Fructose | 180.0 | 800.0 | 4.4444447 |
D-Glucose (Dextrose) | 180.0 | 1400.0 | 7.7777777 |
Fumaric acid | 116.0 | 110.0 | 0.94827586 |
Lactalbumin hydrolysate | | 6660.0 | Infinity |
Malic acid | 134.0 | 1340.0 | 10.0 |
Succinic acid | 118.0 | 120.0 | 1.0169492 |
Sucrose | 342.0 | 53360.0 | 156.02339 |
Yeastolate | | 6660.0 | Infinity |