A solution to accelerate structural elucidation
“To do cryo electron microscopy without native mass spectrometry… is like working with not one hand behind your back, but two hands behind your back…”
Utilize the power of native MS for sample screening
Native mass spectrometry (native MS) is a powerful technique for the characterization of proteins and their complexes. Unlike most common structural biology techniques, which requires a large number of acquisitions for an “average” view of the sample, native MS can reveal all proteoforms and their conformations with just one measurement.
How can native MS help with sample screening?
This software, which analyzes Online Buffer Exchange (OBE)- native MS data, applies novel in-house developed algorithms for larger isotopically unresolved and smaller isotopically resolved protein species. Algorithm and parameters are selected automatically, based on user-defined protein target mass, and run data is processed concurrently on existing instrument hardware so that results are ready shortly after the last run is completed.
Focus on results (instead of the process!)
Thermo Scientific OptiMSe integrates into your workflow seamlessly. It facilitates the preparation of LC queues and tuning of the MS instrument, automates the acquisition and processing of the MS data, and generates an on-the-fly report to aid sample selection for downstream cyro-EM analysis. MS expertise is not required to operate OptiMSe.
Rapid Data Acquisision and analysis
Confidence in Sample Prep
Using native MS to check:
Optimize your EM utilization
“To do cryo electron microscopy without native mass spectrometry… is like working with not one hand behind your back, but two hands behind your back…”
Experts discuss the role of native MS in structural biology and its innovative pairing with cryo-EM
What if you could screen dozens of different buffers or sample preparation conditions in a single day?
With the Thermo Scientific VitroEase buffer optimization kit combined with OptiMSe screening workflow, you no longer need to guess how to best prepare your sample or rely on "good enough."
Enable multiple experiments and sample processing and handling for membrane proteins and complexes
OptiMSe workflow completes each membrane protein measurement within minutes, which enables high-throughput assessment of membrane protein integrity prior to its structural elucidation.
Characterize proteins and composite glycosylation & microheterogeneity for native intact mass analysis
OptiMSe software enables automated data analysis and delivers confidence in results in minutes. Native OBE-1, online buffer exchange column, enables rapid separation at a micro flow rate for direct screening of therapeutic proteins with increased throughput and sensitivity.
Optimize protein overexpression with OBE for nMS applications and see how it can be extended by coupling to affinity separation (e.g., IMAC-OBE).
Perform the highest quality native intact mass and top-down analysis in structural biology and biopharma research with the Q Exactive UHMR mass spectrometer.
Perform high-throughput native mass analysis of protein therapeutics including mAbs with OBE column and Orbitrap Ascend Tribrid MS
* Required field