Our Sterilin line of high-quality, single-use products strengthens our consumables offering for the microbiology, life science, and clinical laboratories.
Featured Sterilin lab consumables categories
The autoclave bags are designed for contaminated waste disposal. The food-grade homogenizer bags are ideal for homogenizing food samples before microbiological analysis.
Leakproof Sterilin containers are used to contain both valuable and hazardous samples which are usually subjected to the rigors of hospital air transport systems.
These include bench guards, weighing boats, loops and spreaders, pipette tips, bulbs and test tube holders, and cones for funnels and flasks.
Both plastic and glass pipettes are ideal for safe and accurate liquid transfer, and suitable for general laboratory use, storage, and transport.
Aseptically manufactured Sterilin Petri Dishes come in a range of sizes, from 30 to 140mm, to suit most applications.
Black 96-well plates are ideal for fluorescence work while the white 96-well plates are ideal for bioluminescence studies.
Sterilin swabs simplify collection, transport, maintenance, and storage of microbiological specimens for clinical and environmental applications.
Sterilin silicone tubing, stoppers, and discs are suitable for use in medical, pharmaceutical and food applications over a temperature range of -50° to +200°C.
Sterilin culture tubes, centrifuge tubes, and scintillation vials are precision manufactured for strength, chemical resistance, and a variety of applications.
Sterilin polystyrene and polyethylene water sampling bottles are the convenient way of sampling both chlorinated and non-chlorinated water for microbiological analysis.
- Need technical support? Contact our expert team for technical and application support of Laboratory Products
California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.