Apply different temperature profiles in your process:
The 40 L/D barrel is segmented in 5 L/D steps. Each segment can be individually heated or cooled, giving you an unmatched precision in applying different temperature profiles to your process.
Process small amount of raw materials:
If a process requires a small amount of raw materials or if there is concern that a material may degrade due to a long residence time, a screw length reduction kit is available.
Segmented screw configurations offer a solution for feeding and mixing of challenging materials such as fibers.