Get fast and highly reliable insight into the safety of indoor air for risk mitigation. Help protect employees and communities with confidence, while maintaining business and operational continuity. Next-level confidence, safety, and reliability.
The AerosolSense Sampler is an in-air pathogen surveillance solution. It is designed to deliver timely and highly reliable insight into the safety of a facility by identifying the presence of in-air pathogens, including Flu A/B, SARS-CoV-2, and/or RSV through indoor air sampling that is sent to a laboratory for PCR testing.
It is recommended to place the AerosolSense Sampler on an unobstructed flat surface like a table that is two to five feet from the ground. Place the sampler in spaces that are high traffic (waiting rooms, employee break rooms) and/or outside containment areas. Since facility spaces vary in size, layout and function, multiple samplers may be needed. Sampler performance has been tested in areas between 150 ft2 - 2,000 ft2.
A 2 hour minimum is recommended for adequate sample collection. Sample collection is flexible to fit a facility’s needs. It is recommended to collect more air samples in high traffic spaces. For further details, visit the support page.
The sampler has been demonstrated to capture particles from 0.1 – 15 um in diameter.
Dust (ranging from ambient dust to high dust load) does not impact the identification or the intensity of identification of aerosolized SARS-CoV-2. This is demonstrated in an evaluation research paper.
The sampler does not use filtration. Particles are trapped on the collection substrate as the air is drawn through the sampler. There is not a degradation of sample performance as the sample gets loaded. Filtration requires more power to ensure the flow of air through the sample and can clog when particles build up, which limits the amount of sampled air.
A green LED indicates that the sampler is in process of collecting an air sample. A yellow LED indicates that the sample cartridge was installed correctly and is ready for sampling. If the door is closed and there is a red LED, that indicates a fault, and also indicates that the sampler is unable to collect a sample. Review the LED chart on the support page.
With an easy-to-use design, any delegated person can operate the sampler with minimal training.
The sampler is pre-calibrated at the manufacturing site. No additional calibration is needed before using the sampler.
The sampler is designed to operate continuously; therefore, you do not need to wait between consecutive sampling.
We recommend using a sample cartridge for no more than 24 hours to ensure users can take timely action after obtaining results.
Sequencing of the virus captured by the sampler for research purposes has been performed by our collaborators. At present, we don’t offer the service.
Yes, if needed an additional label can be added to the sample cartridge. The added label cannot cover the factory barcode; otherwise, sampler operation will be impacted.
Air samples collected on the substrate within the sample cartridge must be sent to a laboratory for PCR testing to determine the presence of RNA from SARS-CoV-2. There are two testing options: Self-testing utilizing an onsite laboratory for ≤ 4h report availability, or the Thermo Fisher Scientific testing service for ≤ 24h report availability.
The AerosolSense Sampler has been validated using the multiplex diagnostic solution Applied Biosystems™ TaqPath™ COVID-19 Combo Kit, Applied Biosystems™ TaqMan™ SARS-CoV-2, FluA, FluB RT-PCR Assay Kit and Applied Biosystems™ TaqMan™ SARS-CoV-2, Flu A/B, RSV RT-PCR Assay Kit.
We provide a sample preparation procedure to elute the sample off of the substrate before starting the PCR workflow. Result interpretations are based on the TaqPath kit Ct cutoff values to determine if sample is Positive, Negative, or Inconclusive.
The box we use for shipping the sample to the partner laboratory (testing service) is identified as UN3373. The biohazard bags for this testing service are 95kpa, which is needed for the UN3373. The sample is isolated within the cartridge (primary), 95kpa biohazard bag (secondary), and then the outer UN3373 box.
The sampler scans the cartridge barcode once inserted. Only one cartridge at a time can be inserted into the sampler. The scan links the cartridge with the sampler ID as well as the sampling start and end time and date. The laboratory will link the test results to the cartridge barcode that is ultimately linked to the specific sampler.
Generally, a positive test result from a sample collected with the AerosolSense Sampler implies that SARS-CoV-2 particles were present in the ambient air surrounding the AerosolSense Sampler during the time of sampling. The sampler has been tested with a 0.34 genome copies per liter of air limit of detection. As such you should follow your established OSHA, EHS, local, facility, and/or SARS-CoV-2 safety protocols to ensure you limit the risk of exposure and transmission within your facility.
The AerosolSense Sampler is an in-air pathogen surveillance solution. It is designed to deliver timely and highly reliable insight into the safety of a facility by identifying the presence of in-air pathogens, including Flu A/B, SARS-CoV-2, and/or RSV through indoor air sampling that is sent to a laboratory for PCR testing.
It is recommended to place the AerosolSense Sampler on an unobstructed flat surface like a table that is two to five feet from the ground. Place the sampler in spaces that are high traffic (waiting rooms, employee break rooms) and/or outside containment areas. Since facility spaces vary in size, layout and function, multiple samplers may be needed. Sampler performance has been tested in areas between 150 ft2 - 2,000 ft2.
A 2 hour minimum is recommended for adequate sample collection. Sample collection is flexible to fit a facility’s needs. It is recommended to collect more air samples in high traffic spaces. For further details, visit the support page.
The sampler has been demonstrated to capture particles from 0.1 – 15 um in diameter.
Dust (ranging from ambient dust to high dust load) does not impact the identification or the intensity of identification of aerosolized SARS-CoV-2. This is demonstrated in an evaluation research paper.
The sampler does not use filtration. Particles are trapped on the collection substrate as the air is drawn through the sampler. There is not a degradation of sample performance as the sample gets loaded. Filtration requires more power to ensure the flow of air through the sample and can clog when particles build up, which limits the amount of sampled air.
A green LED indicates that the sampler is in process of collecting an air sample. A yellow LED indicates that the sample cartridge was installed correctly and is ready for sampling. If the door is closed and there is a red LED, that indicates a fault, and also indicates that the sampler is unable to collect a sample. Review the LED chart on the support page.
With an easy-to-use design, any delegated person can operate the sampler with minimal training.
The sampler is pre-calibrated at the manufacturing site. No additional calibration is needed before using the sampler.
The sampler is designed to operate continuously; therefore, you do not need to wait between consecutive sampling.
We recommend using a sample cartridge for no more than 24 hours to ensure users can take timely action after obtaining results.
Sequencing of the virus captured by the sampler for research purposes has been performed by our collaborators. At present, we don’t offer the service.
Yes, if needed an additional label can be added to the sample cartridge. The added label cannot cover the factory barcode; otherwise, sampler operation will be impacted.
Air samples collected on the substrate within the sample cartridge must be sent to a laboratory for PCR testing to determine the presence of RNA from SARS-CoV-2. There are two testing options: Self-testing utilizing an onsite laboratory for ≤ 4h report availability, or the Thermo Fisher Scientific testing service for ≤ 24h report availability.
The AerosolSense Sampler has been validated using the multiplex diagnostic solution Applied Biosystems™ TaqPath™ COVID-19 Combo Kit, Applied Biosystems™ TaqMan™ SARS-CoV-2, FluA, FluB RT-PCR Assay Kit and Applied Biosystems™ TaqMan™ SARS-CoV-2, Flu A/B, RSV RT-PCR Assay Kit.
We provide a sample preparation procedure to elute the sample off of the substrate before starting the PCR workflow. Result interpretations are based on the TaqPath kit Ct cutoff values to determine if sample is Positive, Negative, or Inconclusive.
The box we use for shipping the sample to the partner laboratory (testing service) is identified as UN3373. The biohazard bags for this testing service are 95kpa, which is needed for the UN3373. The sample is isolated within the cartridge (primary), 95kpa biohazard bag (secondary), and then the outer UN3373 box.
The sampler scans the cartridge barcode once inserted. Only one cartridge at a time can be inserted into the sampler. The scan links the cartridge with the sampler ID as well as the sampling start and end time and date. The laboratory will link the test results to the cartridge barcode that is ultimately linked to the specific sampler.
Generally, a positive test result from a sample collected with the AerosolSense Sampler implies that SARS-CoV-2 particles were present in the ambient air surrounding the AerosolSense Sampler during the time of sampling. The sampler has been tested with a 0.34 genome copies per liter of air limit of detection. As such you should follow your established OSHA, EHS, local, facility, and/or SARS-CoV-2 safety protocols to ensure you limit the risk of exposure and transmission within your facility.
Need further support? Visit the In-Air Pathogen Surveillance Support page ›
Every day there’s a new headline about the best ways to test for SARS-CoV-2 virus, the virus that causes COVID-19. In this Q&A, Joshua Trotta, Senior Director Business Development, discusses what you need to know about qPCR, the gold standard for COVID-19 testing. This is a follow-up to our “How does a COVID-19 test work?” interview with Joshua on Science with a Twist earlier this year.
New information is rapidly emerging, and scientists are working to learn more about these variants to better understand how easily they might be transmitted, whether they may confer increased virulence, the effectiveness of currently authorized vaccines against them, and more. Additionally, COVID-19 testing labs may have questions about the potential impact of the mutations on current molecular diagnostic tests.