Advanced PCR-based COVID-19 test design for NPS and nasal swabs using RNase P, to detect infections in symptomatic and asymptomatic patients
Features of the TaqPath COVID-19 RNase P Combo Kit 2.0:
- Human sample confirmation—the RNase P gene serves as an endogenous control to help ensure sample integrity, quality, and extraction
- Innovative, multi-target assay design—multiple targets on orf1a, orf1b, and N genes compensate for emerging SARS-CoV-2 mutations, helping provide increased confidence in results
- Sensitive detection for symptomatic or asymptomatic cases—PCR-based test detects active infections from individuals suspected of having COVID-19 by their health care provider as well as asymptomatic individuals; 75 GCE/mL LOD
- Affordable and scalable—add to your existing COVID-19 test options using some of the most widely used PCR systems in the world
Standard workflow
The TaqPath COVID-19 RNase P Combo Kit 2.0 utilizes a standard workflow that provides results in ~3 hours.
Receive samples | Prepare samples | Prepare plate and run PCR | Analyze and report |
Nasopharyngeal swabs |
Thermo Scientific KingFisher Flex Purification System with Applied Biosystems MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit
|
| Applied Biosystems Pathogen Interpretive Software (v1.1.0) |
Find your IFU
Find the approved Instructions for Use (IFU) for your country. (Note: Product availability varies by country.)
Ordering information
Product | Components included | Qty | Cat. No. |
---|---|---|---|
TaqPath COVID-19 RNase P Assay 2.0 (1 tube) | 1,000 rxn | ||
TaqPath COVID-19 Plus Control (10 tubes) | |||
TaqPath Control Dilution Buffer (1 box with 10 tubes) | |||
TaqPath 1-Step Multiplex Master Mix, No ROX (1 bottle with 10 mL) (required, but not included in kit) |
Simple analysis and reporting
Applied Biosystems Pathogen Interpretive Software (v1.1.0) Edition is part of the required workflow and must be downloaded prior to instrumentation use.
Pathogen Interpretive Software helps your lab decrease analysis and interpretation time and reduce risk of user interpretation error. You may download the software upon kit purchase. Labs following the authorized workflow are required to complete an eLearning course and pass an exam to download.
Follow the instructions below to obtain the software from your local support team:
- Go to thermofisher.com/contactus
- Select “Technical Support” as category
- Enter product name or SKU and select your country
- Call the local number displayed on the screen
For In Vitro Diagnostic Use. For Emergency Use Authorization (EUA) Only | Rx Only
© 2021 Thermo Fisher Scientific, Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.