Live HeLa cells stained with the reagents in the Image-iT® LIVE Lysosomal and Nuclear Labeling Kit.
Live HeLa cells were transfected using pShooter vector pCMV/myc/mito/GFP and Lipofectamine 2000 transfection reagent and stained with the reagents in the Image-iT® LIVE Lysosomal and Nuclear Labeling Kit (I34202). Lysosomes were stained with LysoTracker® Red DND-99. Cells were visualized using epifluorescence microscopy.
Related Products
Related Images
Live cell imaging with CellLight™ reagents. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Bacteriophage T4 replication proteins stained with SYPRO® Orange Protein Gel Stain. Go ›
Madin-Darby canine kidney cells. LysoSensor Yellow/Blue DND-160. Go ›
Imaging autophagy in live HeLa cells with CellLight® reagents for mitochondria and lysosomes: Go ›
Porcine primary skeletal muscle cell transduced with Organelle Lights fluorescent proteins. Go ›