Endogenous phosphatase activity in a zebrafish ovary cross section.
Endogenous phosphatase activity in a zebrafish ovary cross-section, stained with the BCIP (B6492) and NBT (N6495) reagents. These reagents are also available in the NBT/BCIP Reagent Kit (N6547).