Simultaneous detection of three gene targets in a whole-mount Drosophila embryo by fluorescence in situ hybridization.
Pseudocolored green-fluorescent labeling represents a fluorescein-labeled cRNA probe detected using a rabbit anti-fluorescein/Oregon Green® primary antibody (Cat. No. A889) and an Alexa Fluor® 488 dye–labeled anti-rabbit secondary antibody (Cat. No. A11008). Pseudocolored yellow- and red-fluorescent labeling represents a biotinylated cRNA probe detected using HRP–streptavidin and Alexa Fluor® 568 tyramide (TSA Kit #24, Cat. No. T20934). Pseudocolored blue-fluorescent labeling represents a digoxigenin-labeled cRNA probe detected using a mouse anti-digoxigenin primary antibody in conjunction with an Alexa Fluor® 647 dye–labeled anti-mouse secondary antibody (Cat. No. A21235). The image was contributed by Ethan Bier and David Kosman, University of California, San Diego.
Related Products
Related Images
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Labeled paint probes hybridized to human metaphase chromosomes Go ›
Immunocytochemistry using Alpha-Tubulin Monoclonal Antibody, Mouse Go ›
Tyramide signal amplification of immunofluorescent staining in mouse brain sections. Go ›