Now available to order online!
384-well microfluidic cards enable easy and affordable profiling and/or validation of hundreds of targets. TaqMan® Array Cards can be customized using any predesigned assays pre-spotted in the wells. Minimum order quantity of ten cards.
Click here for Custom TaqMan® Array Advanced miRNA Cards