Broad range of reliable and simple PCR-based testing to help diagnose infections
Prompt and early diagnosis of sexually transmitted infections (STIs) is important to enable appropriate treatment and help prevent further transmission. Clinical testing labs require STI testing solutions that help provide confidence, fast turnaround time, and accurate results. The TaqPath Menu STI portfolio supports sexual health testing with CE-marked sexually transmitted infection (STI) assays, designed to provide accurate, reliable results with excellent sensitivity and specificity across a wide variety of bacteria (such as Chlamydia trachomatis (CT), Neisseria gonorrhoeae (NG) and mycoplasma genitalium), parasites (such as Trichomonas vaginalis), or viruses (such as human papillomavirus (HPV) or Herpes simplex virus (HSV-1/2)). This helps give confidence in screening, diagnostic, and treatment decisions, and helps stop the spread of disease.
Rely on a comprehensive portfolio of TaqPath Menu kits to meet your lab’s clinical testing needs 
  • Extensive solution for STI with multiplex kits
  • Simple laboratory workflow
  • High sensitivity and specificity
  • Contamination reduction
  • Compatible with a wide range of real-time PCR devices
Free Webinar: Molecular diagnostic approaches to accelerate and improve STI diagnosis
Learn about the importance STI testing and the TaqPath™ Menu STI portfolio
Want to use our solutions?  Contact us for details on our limited time offer.
More information from one reaction
  • Detects and distinguishes STI pathogens in a single reaction tube:
    • Chlamydia trachomatis
    • Neisseria gonorrhoeae
    • Mycoplasma genitalium
Simple laboratory workflow
  • Easily combinable with other PCR kits from the TaqPath Menu in one workflow
Easy-to-use concept
  • Single tube Ready-to-Use Master Mix contains all components for PCR amplification
  • No additional PCR reagent pipetting necessary
Download flyer ›
High sensitivity
  • Detects porA mutant of Neisseria gonorrhoeae and Swedish variant of Chlamydia trachomatis
  • Secured by targeting the multicopy sequence of 16S rRNA for all three detected pathogens
Contamination reduction
  • Master Mix contains Uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
Compatible with a wide range of real-time PCR devices
CE-IVD certified diagnostic test
High specificity
  • Unique detection of 24 high-risk HPV types
Differentiation of high-risk types 16, 18, and 45
  • HPV16 and HPV18 are responsible for 70% of cervical cancer cases and pre-cancerous cervical lesions
  • HPV45 is the third most prevalent genotype in cases of invasive cervical cancer worldwide (5–6%) 
  • Enables specific management of patient monitoring and therapy
Self-sampling option
  • Supports scale-up of HPV screening through simple and comfortable sampling 
  • Housekeeping GAPDH allows for correct sampling control
Download flyer ›
Supports compliance with guidelines for HPV screening
  • German S3-guidelines “Prevention of cervical cancer” (S3-Leitlinie Prävention des Zervixkarzinoms Langverison 1.0 – Dezember 2017)
  • The clinical and reproducibility criteria of the International Guidelines for HPV tests for use in cervical cancer screening
Compatible with a wide range of real-time PCR devices
CE-IVD diagnostic test
More information from one reaction
  • Detects and distinguishes STI pathogens in a single reaction tube:
    • Mycoplasma hominis
    • Ureaplasma urealyticum
    • Ureaplasma parvum
Simple laboratory workflow
  • Easily combinable with other PCR kits from the TaqPath Menu in one workflow
Contamination reduction
  • Ready-to-Use Master Mix contains Uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
Download flyer ›
Easy-to-use concept
  • Single tube Ready-to-Use Master Mix contains all components for PCR amplification
  • No additional PCR reagent pipetting necessary
Compatible with a wide range of real-time PCR devices
CE-IVD diagnostic test
High specificity
  • Secured by a multicopy sequence of the beta-tubulin gene
  • Excellent sensitivity, reaching up to 0.25 cp/μL
Reliable detection
  • Simple sensitive detection compared to wet mount microscopy and culture
Easy-to-use concept
  • Single tube Ready-to-Use Master Mix contains all components for PCR amplification
  • No additional PCR reagent pipetting necessary
Download flyer ›
Contamination reduction
  • Master Mix contains Uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
Simple laboratory workflow
  • Easily combinable with other TaqPath Menu PCR kits in one workflow
Compatible with a wide range of real-time PCR devices
CE-IVD diagnostic test
More information from one reaction
  • PCR kit detects and distinguishes Herpes Simplex Virus 1 and Herpes Simplex Virus 2 in a single reaction tube
Excellent quality
  • The highest possible diagnostic sensitivity (100.00%) and diagnostic specificity (97.62%)
Simple laboratory workflow
  • Easily combinable with other PCR kits from the TaqPath Menu in one workflow
Download flyer ›
Easy-to-use concept
  • Single tube Ready-to-Use Master Mix contains all components for PCR amplification
  • No additional PCR reagent pipetting necessary
Contamination reduction
  • Ready-to-Use Master Mix contains Uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
Compatible with a wide range of real-time PCR devices
CE-IVD certified diagnostic test
High sensitivity
  • Secured by targeting the multicopy sequence of the 16S rRNA gene
  • Dual target detection of the 16S rRNA gene and a cryptic plasmid
Simple laboratory workflow
  • Easily combinable with other PCR kits from the TaqPath Menu in one workflow
Easy-to-use concept
  • Single tube Ready-to-Use Master Mix contains all components for PCR amplification
  • No additional PCR reagent pipetting necessary
Download flyer ›
Reliable detection of Swedish variant of Chlamydia trachomatis
  • Reduction of false-negative results caused by a sequence variation in the cryptic plasmid
Contamination reduction
  • Master Mix contains Uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
Compatible with a wide range of real-time PCR devices
CE-IVD diagnostic test
Dual target detection
  • Dual target detection of the 16S rRNA gene and the porA pseudogene
High sensitivity
  • Secured by targeting the multicopy sequence of the 16S rRNA gene
Reliable detection of porA mutant
  • Reduction of false-negative results caused by a sequence variation in the porA pseudogene
Download flyer › 
Contamination reduction
  • Master Mix contains Uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
Compatible with a wide range of real-time PCR devices
CE-IVD diagnostic test
More information with less samples
  • Helps distinguish Mycoplasma hominis from Mycoplasma genitalium using 3-channel system
Simple laboratory workflow
  • Easily combinable with other PCR kits from the TaqPath Menu in one workflow
High sensitivity
  • Secured by targeting the multicopy sequence of the 16S rRNA gene of Mycoplasma genitalium
Download flyer ›
Contamination reduction
  • Master Mix contains Uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
Compatible with a wide range of real-time PCR devices
CE-IVD diagnostic test
Comprehensive solution for STI
  • As part of an STI Panel, enables a comprehensive solution for symptomatic STI diagnostics
  • Suitable especial for differential diagnostics of non-candidal vulvovaginitis
Simple laboratory workflow
  • Easily combinable with other PCR kits from the TaqPath Menu in one workflow
Easy-to-use concept
  • Single tube Ready-to-Use Master Mix contains all components for PCR amplification
  • No additional PCR reagent pipetting necessary
Download flyer ›
Contamination reduction
  • Master Mix contains Uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
Compatible with a wide range of real-time PCR devices
CE-IVD diagnostic test
More information with less samples
  • Enables the identification of Ureaplasma urealyticum using 3-channel system
Simple laboratory workflow
  • Easily combinable with other PCR kits from the TaqPath Menu in one workflow
Contamination reduction
  • Master Mix contains Uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
Download flyer ›
Compatible with a wide range of real-time PCR devices
CE-IVD diagnostic test
Highly specific detection
  • Secured by highly conservative polA gene for DNA polymerase I
Easy-to-use concept
  • Single tube Ready-to-Use Master Mix contains all components for PCR amplification
  • No additional PCR reagent pipetting necessary
Contamination reduction
  • Master Mix contains Uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
Download flyer › 
Simple laboratory workflow
  • Easily combinable with other TaqPath Menu | GeneProof PCR kits in one workflow
Compatible with a wide range of real-time PCR devices
CE-IVD diagnostic test
Order information
Resources
For In Vitro Diagnostic Use.
Intended use and regulatory statements of products vary. For specific intended use and regulatory statements please refer to the Instructions for Use (IFU). Product availability by country varies. For product availability in your country, check the associated product webpages.
TaqPath™ Menu | GeneProof® assays are manufactured by GeneProof® and distributed by Thermo Fisher Scientific
© 2023 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries. GeneProof® is the property of GeneProof.
Style Sheet for Global Design System