Request a Quote
B-27 Serum-Free Supplement (50X) liquid
B-27 Serum-Free Supplement is used for growth and long-term viability of hippocampal neurons.
This supplement is to be used with a combination of basal medium such as Neurobasal Medium. Below is the composition list. The concentrations are proprietary.
Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Vitamins | |||
Biotin | confidential | n/a | |
DL Alpha Tocopherol Acetate | confidential | n/a | |
DL Alpha-Tocopherol | confidential | n/a | |
Vitamin A (acetate) | confidential | n/a | |
Proteins | |||
BSA, fatty acid free Fraction V | confidential | n/a | |
Catalase | confidential | n/a | |
Human Recombinant Insulin | confidential | n/a | |
Human Transferrin | confidential | n/a | |
Superoxide Dismutase | confidential | n/a | |
Other Components | |||
Corticosterone | confidential | n/a | |
D-Galactose | confidential | n/a | |
Ethanolamine HCl | confidential | n/a | |
Glutathione (reduced) | confidential | n/a | |
L-Carnitine HCl | confidential | n/a | |
Linoleic Acid | confidential | n/a | |
Linolenic Acid | confidential | n/a | |
Progesterone | confidential | n/a | |
Putrescine 2HCl | confidential | n/a | |
Sodium Selenite | confidential | n/a | |
T3 (triodo-I-thyronine) | confidential | n/a |
Reference:
Related Products:
B-27 Supplement Minus AO (50X) liquid
Cat No:
10889038
B-27 Supplement Minus Vitamin A (50X) liquid
Cat No:
12587001
,
12587010