Detection of oxidative stress in live cells using the Image-iT® LIVE Reactive Oxygen Species (ROS) Kit.
Detection of oxidative stress in live cells using the Image-iT® LIVE Reactive Oxygen Species (ROS) Kit (I36007). Live bovine pulmonary artery endothelial cells were treated with tert-butyl hydroperoxide to induce oxidative stress (bottom) or were left untreated (top). Cells were then labeled with carboxy-H2DCFDA, which fluoresces when oxidized by ROS, and nuclei were stained with blue-fluorescent Hoechst 33342. The stressed cells exhibited green fluorescence, signaling an increase in ROS, whereas the untreated cells showed minimal fluorescence.
Related Products
Related Images
Oxidative stress analysis in BPAE and Raw cells using CellROX® Deep Red Reagent (Cat. No. C10422) Go ›
BPAE detection of oxidative stress using MitoTracker® Green (Cat. No. M7514) and CellROX® Deep Red Reagent (Cat. No. C10422) Go ›
Agarose gel containing camptothecin-treated HL-60 cells. Go ›
Imaging autophagy in live HeLa cells with CellLight® reagents for mitochondria and lysosomes: Go ›
Live cell imaging with CellLight™ reagents. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›