Immunohistochemistry using GFAP Monoclonal Antibody, Mouse, Alexa Fluor® 594 Conjugate:
Immunohistochemistry using GFAP Monoclonal Antibody, Mouse Alexa Fluor® 594 Conjugate: Rat brain cryosections were labeled with the Alexa Fluor® 594 conjugate of anti–glial fibrillary acidic protein antibody. Rat brain cryosections were labeled with the red-fluorescent Alexa Fluor® 594 conjugate of anti–glial fibrillary acidic protein antibody (Cat. no. A21295). Nuclei were counterstained with TOTO®-3 iodide (Cat. no. T3604, pseudocolored blue).
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›