Bovine pulmonary artery endothelial cells.
Live bovine pulmonary artery endothelial cells stained with ER-Tracker™ Blue-White DPX (Cat. No. E12353) and MitoTracker Red CM-H2XRos (Cat. No. M7513). The endoplasmic reticulum appears green and the mitochondria appear orange. The image was acquired using a fluorescence microscope equipped with a triple-bandpass filter set appropriate for DAPI, fluorescein and Texas Red® dyes.
Related Products
Related Images
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Immunofluorescent staining using mouse anti-Golgin-97 monoclonal antibody Go ›