Kangaroo rat (PtK2) cells. LIVE/DEAD® Viability/Cytotoxicity Kit.
Live and dead kangaroo rat (PtK2) cells stained with ethidium homodimer-1 and the esterase substrate calcein AM, both of which are provided in our LIVE/DEAD® Viability/Cytotoxicity Kit (L3224). Live cells fluoresce a bright green, whereas dead cells with compromised membranes fluoresce red-orange.
Related Products
Related Images
Live cell imaging with CellLight™ reagents. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
BPAE cells fixed and permeabilized using the Image-iT® Fixation/Permeabilization Kit. Go ›
BPAE cells fixed and permeabilized using the Image-iT® Fixation/Permeabilization Kit. Go ›