Bacteriophage T4 replication proteins stained with SYPRO® Orange Protein Gel Stain.
Purified bacteriophage T4 replication proteins that have been electrophoresed on a 12% SDS-polyacrylamide gel and then stained with SYPRO® Orange protein gel stain (Cat. no. S6650, S6651). Gel staining was visualized with 300 nm epi-illumination. T4 phage proteins were a gift from the laboratory of Peter von Hippel, Institute of Molecular Biology, University of Oregon.
Related Products
Related Images
2D gel stained with the SYPRO Ruby protein gel stain and the Pro-Q Emerald 300 reagent. Go ›
Comparison of two different 2D protein gels stained with SYPRO® Ruby protein gel stain. Go ›
Live cell imaging with CellLight™ reagents. Go ›
Madin-Darby canine kidney cells. LysoSensor Yellow/Blue DND-160. Go ›