Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012463.hg.1 |
ENST00000500112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012463.hg.1 |
ENST00000500112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012463.hg.1 |
ENST00000500112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012463.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012463.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012463.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012463.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012463.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188113.hg.1 |
ENST00000500112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188113.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188113.hg.1 |
ENST00000500112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188113.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188113.hg.1 |
TC0800012463.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800188111.hg.1 |
ENST00000500112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188111.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188111.hg.1 |
ENST00000500112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188111.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188111.hg.1 |
TC0800012463.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800188114.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188114.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188114.hg.1 |
TC0800012463.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800012463.hg.1 |
ENST00000500112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012463.hg.1 |
ENST00000500112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012463.hg.1 |
ENST00000500112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012463.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012463.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012463.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012463.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012463.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188112.hg.1 |
ENST00000500112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188112.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188112.hg.1 |
ENST00000500112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188112.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188112.hg.1 |
TC0800012463.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800188110.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188110.hg.1 |
NR_108049.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188110.hg.1 |
TC0800012463.hg.1 |
Clariom D TC |