Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012044.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012044.hg.1 |
TC14001056.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000250448.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000250448.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000250448.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000250448.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000250448.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000250448.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000545425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000545425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000545425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000545425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000545425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000553751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000553751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000553751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000553751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000553751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000554607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000554607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000554607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000554607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000554607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000557418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000557418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
ENST00000557418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008981.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400166124.hg.1 |
ENST00000250448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166124.hg.1 |
ENST00000553751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166124.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400166124.hg.1 |
ENST00000250448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166124.hg.1 |
ENST00000553751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166124.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400166124.hg.1 |
TC1400008981.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400166127.hg.1 |
ENST00000553751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166127.hg.1 |
ENST00000557418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166127.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400166127.hg.1 |
ENST00000553751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166127.hg.1 |
ENST00000557418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166127.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400166127.hg.1 |
TC1400008981.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400166122.hg.1 |
ENST00000250448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166122.hg.1 |
ENST00000553751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166122.hg.1 |
ENST00000554607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166122.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400166122.hg.1 |
ENST00000250448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166122.hg.1 |
ENST00000553751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166122.hg.1 |
ENST00000554607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166122.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400166122.hg.1 |
TC1400008981.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000250448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000250448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000250448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000250448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000250448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000250448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000545425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000545425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000545425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000545425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000545425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000553751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000553751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000553751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000553751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000553751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000554607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000554607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000554607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000554607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000554607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000557418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000557418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
ENST00000557418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
NM_004496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008981.hg.1 |
NM_004496.3 |
Refseq |