Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024690.hg.1 |
ENST00000424202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024690.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024690.hg.1 |
TC12001890.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000392905.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000392905.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000392905.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000392905.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000424202.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000424202.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000424202.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000424202.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000424202.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000481539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000481539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000481539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000481539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
ENST00000481539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024688.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024688.hg.1 |
ENST00000337514.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024688.hg.1 |
ENST00000392904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024688.hg.1 |
ENST00000424202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024688.hg.1 |
ENST00000456098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024688.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024688.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024688.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024688.hg.1 |
TC12001890.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000392905.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000392905.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000392905.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000392905.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000424202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000424202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000424202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000424202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000424202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000481539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000481539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000481539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000481539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
ENST00000481539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011727.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190862.hg.1 |
ENST00000424202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190862.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190862.hg.1 |
ENST00000424202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190862.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190862.hg.1 |
TC1200011727.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
ENST00000392905.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
ENST00000424202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
ENST00000307046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
ENST00000337514.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
ENST00000392904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
ENST00000392905.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
ENST00000424202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
ENST00000456098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
NM_000618.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
NM_001111283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
NM_001111284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190859.hg.1 |
TC1200011727.hg.1 |
Clariom D TC |