Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000461614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000461614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000461614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000462805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000462805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000462805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000480202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000480202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000480202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000480202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
ENST00000480202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000461614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000461614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000461614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000462805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000462805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000462805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000480202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000480202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000480202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000480202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
ENST00000480202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013028.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196018.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196018.hg.1 |
ENST00000480202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196018.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196018.hg.1 |
ENST00000358432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196018.hg.1 |
ENST00000480202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196018.hg.1 |
NM_004431.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196018.hg.1 |
TC0100013028.hg.1 |
Clariom D TC |