Assay Details
Gene Symbol: CCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123396.1 | NP_001116868.1 | 2 | 1957 | 61 | |
XM_011534069.1 | XP_011532371.1 | 3 | 1670 | 61 | ||
GenBank mRNA | DA927445.1 | - | 1 | 199 | 61 | |
U03905.1 | - | 2 | 1552 | 61 | ||
X95583.1 | - | 1 | 812 | 61 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0300007256.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000255.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000255.hg.1
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel remodeling
dendritic cell chemotaxis
inflammatory response to antigenic stimulus
positive regulation of T-helper 1 type immune response
negative regulation of type 2 immune response
cellular calcium ion homeostasis
chemotaxis
inflammatory response
immune response
cellular defense response
G-protein coupled receptor signaling pathway
negative regulation of adenylate cyclase activity
positive regulation of cytosolic calcium ion concentration
JAK-STAT cascade
aging
response to wounding
response to ionizing radiation
regulation of vascular endothelial growth factor production
positive regulation of gene expression
positive regulation of T cell chemotaxis
negative regulation of angiogenesis
cytokine-mediated signaling pathway
cellular homeostasis
negative regulation of cell migration
negative regulation of axon extension
forebrain development
positive regulation of fever generation
response to lipopolysaccharide
positive regulation of interferon-gamma production
positive regulation of interleukin-2 production
positive regulation of interleukin-6 production
T-helper 17 cell chemotaxis
positive regulation of tumor necrosis factor biosynthetic process
defense response to bacterium
negative regulation of eosinophil degranulation
fat cell differentiation
positive regulation of neuron differentiation
positive regulation of alpha-beta T cell proliferation
positive regulation of interleukin-1 beta secretion
positive regulation of inflammatory response
positive regulation of T cell activation
positive regulation of apoptotic process by virus
chemokine-mediated signaling pathway
neuron death
positive regulation of monocyte chemotaxis
positive regulation of cell migration involved in sprouting angiogenesis
positive regulation of immune complex clearance by monocytes and macrophages
negative regulation of neural precursor cell proliferation
positive regulation of monocyte extravasation
positive regulation of CD8-positive, alpha-beta T cell extravasation
positive regulation of astrocyte chemotaxis
positive regulation of hematopoietic stem cell migration
chemokine receptor activity
C-C chemokine receptor activity
protein kinase binding
CCR2 chemokine receptor binding
protein homodimerization activity
chemokine (C-C motif) ligand 5 binding
blood vessel remodeling
dendritic cell chemotaxis
inflammatory response to antigenic stimulus
positive regulation of T-helper 1 type immune response
negative regulation of type 2 immune response
cellular calcium ion homeostasis
chemotaxis
inflammatory response
immune response
cellular defense response
G-protein coupled receptor signaling pathway
negative regulation of adenylate cyclase activity
positive regulation of cytosolic calcium ion concentration
JAK-STAT cascade
aging
response to wounding
response to ionizing radiation
regulation of vascular endothelial growth factor production
positive regulation of gene expression
positive regulation of T cell chemotaxis
negative regulation of angiogenesis
cytokine-mediated signaling pathway
cellular homeostasis
negative regulation of cell migration
negative regulation of axon extension
forebrain development
positive regulation of fever generation
response to lipopolysaccharide
positive regulation of interferon-gamma production
positive regulation of interleukin-2 production
positive regulation of interleukin-6 production
T-helper 17 cell chemotaxis
positive regulation of tumor necrosis factor biosynthetic process
defense response to bacterium
negative regulation of eosinophil degranulation
fat cell differentiation
positive regulation of neuron differentiation
positive regulation of alpha-beta T cell proliferation
positive regulation of interleukin-1 beta secretion
positive regulation of inflammatory response
positive regulation of T cell activation
positive regulation of apoptotic process by virus
chemokine-mediated signaling pathway
neuron death
positive regulation of monocyte chemotaxis
positive regulation of cell migration involved in sprouting angiogenesis
positive regulation of immune complex clearance by monocytes and macrophages
negative regulation of neural precursor cell proliferation
positive regulation of monocyte extravasation
positive regulation of CD8-positive, alpha-beta T cell extravasation
positive regulation of astrocyte chemotaxis
positive regulation of hematopoietic stem cell migration
chemokine receptor activity
C-C chemokine receptor activity
protein kinase binding
CCR2 chemokine receptor binding
protein homodimerization activity
chemokine (C-C motif) ligand 5 binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03005190.hg.1 | TC03000255.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03005190.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005190.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000400888.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000400888.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000400888.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000400888.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000400888.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000400888.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000400888.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000400888.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000421659.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000421659.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000421659.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000421659.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000465202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000465202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000465202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000465202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | ENST00000465202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03005188.hg.1 | TC03000255.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03005188.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005188.hg.1 | ENST00000465202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005188.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03005189.hg.1 | TC03000255.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03005189.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005189.hg.1 | ENST00000465202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03005189.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300151409.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300151409.hg.1 | ENST00000465202.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300151409.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300151409.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300151409.hg.1 | ENST00000465202.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300151409.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300151409.hg.1 | TC0300007256.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300151410.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300151410.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300151410.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300151410.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300151410.hg.1 | TC0300007256.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300151408.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300151408.hg.1 | ENST00000465202.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300151408.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300151408.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300151408.hg.1 | ENST00000465202.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300151408.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300151408.hg.1 | TC0300007256.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000292301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000400888.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000400888.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000400888.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000400888.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000400888.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000400888.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000400888.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000400888.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000421659.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000421659.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000421659.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000421659.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000445132.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000465202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000465202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000465202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000465202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | ENST00000465202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123041.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007256.hg.1 | NM_001123396.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 110071 | - | - |