Assay Details
Gene Symbol: CDKN1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000389.4 | NP_000380.1 | 2 - 3 | 566 | 66 | |
NM_001220777.1 | NP_001207706.1 | 2 - 3 | 511 | 66 | ||
NM_001220778.1 | NP_001207707.1 | 2 - 3 | 676 | 66 | ||
NM_001291549.1 | NP_001278478.1 | 3 - 4 | 718 | 66 | ||
NM_078467.2 | NP_510867.1 | 3 - 4 | 513 | 66 | ||
GenBank mRNA | AB209881.1 | - | 2 - 3 | 523 | 66 | |
AB451290.1 | - | 1 - 2 | 441 | 66 | ||
AB451422.1 | - | 1 - 2 | 441 | 66 | ||
AK298901.1 | - | 2 - 3 | 575 | 66 | ||
AK309512.1 | - | 3 - 4 | 412 | 66 | ||
BC000275.1 | - | 2 - 3 | 506 | 3355833 | 66 | |
BC000312.2 | - | 2 - 3 | 660 | 2821049 | 66 | |
BC001935.1 | - | 2 - 3 | 521 | 2823668 | 66 | |
BC013967.2 | - | 2 - 3 | 524 | 2822909 | 66 | |
BT006719.1 | - | 1 - 2 | 441 | 66 | ||
BU154329.1 | - | 3 - 4 | 534 | 66 | ||
BU193165.1 | - | 3 - 4 | 718 | 66 | ||
CR536533.1 | - | 1 - 2 | 441 | 66 | ||
DA582789.1 | - | 2 - 3 | 567 | 66 | ||
DA838987.1 | - | 2 - 3 | 474 | 66 | ||
L25610.1 | - | 2 - 3 | 519 | 66 | ||
L26165.1 | - | 2 - 3 | 519 | 66 | ||
L47232.1 | - | 2 - 3 | 494 | 66 | ||
L47233.1 | - | 2 - 3 | 494 | 66 | ||
S67388.1 | - | 2 - 3 | 516 | 66 | ||
U03106.1 | - | 2 - 3 | 516 | 66 | ||
U09579.1 | - | 2 - 3 | 535 | 66 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0600007847.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06000532.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06000532.hg.1
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Endogenous Controls Recommended Assay Homo sapiens Human Signal Transduction Pathways Human p53 Signaling HUMAN COLORECTAL CANCER METASTASIS HUMAN CYCLINS AND CELL CYCLE REGULATION HUMAN MOLECULAR MECHANISMS OF CANCER HUMAN ENDOGENOUS CONTROL FAST PLATE HUMAN BREAST CANCER REGULATION BY STATHMIN1 HUMAN PI3K SIGNALING HUMAN PANCREATIC ADENOCARCINOMA Inventoried MicroArray Quality Control (MAQC) Human_Signal_Transduction_Pathways Human_p53_Signaling Human_Pancreatic_Adenocarcinoma Human_Endogenous_Controls Human_Molecular_Mechanisms_of_Cancer Human_Cyclins_and_Cell_Cycle_Regulation Human_Colorectal_Cancer_Metastasis Human_PI3K_Signaling Human_Breast_Cancer_Regulation_by_Stathmin1 Probe spans exons |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (49)
Down-regulation of serum/glucocorticoid regulated kinase 1 in colorectal tumours is largely independent of promoter hypermethylation.
- Authors
- Lessi F, Beggs A, de Palo M, Anti M, Macarone Palmieri R, Francesconi S, Gomes V, Bevilacqua G, Tomlinson I, Segditsas S
- Journal
- Plos One - Page No e13840 (Fri Jan 01 00:00:00 EST 2010)
- Volume (Year)
- 5 (2010)
- Product Usage
- -
Discovery and optimization of chromenotriazolopyrimidines as potent inhibitors of the mouse double minute 2-tumor protein 53 protein-protein interaction.
- Authors
- Allen JG, Bourbeau MP, Wohlhieter GE, Bartberger MD, Michelsen K, Hungate R, Gadwood RC, Gaston RD, Evans B, Mann LW, Matison ME, Schneider S, Huang X, Yu D, Andrews PS, Reichelt A, Long AM, Yakowec P, Yang EY, Lee TA, Oliner JD
- Journal
- J Med Chem - Page No 7044 (Thu Nov 26 00:00:00 EST 2009)
- Volume (Year)
- 52 (2009)
- Product Usage
- -
Ergosterol peroxide from an edible mushroom suppresses inflammatory responses in RAW264.7 macrophages and growth of HT29 colon adenocarcinoma cells.
- Authors
- Kobori M, Yoshida M, Ohnishi-Kameyama M, Shinmoto H
- Journal
- Br J Pharmacol - Page No 209 (Mon Jan 01 00:00:00 EST 2007)
- Volume (Year)
- 150 (2007)
- Product Usage
- -
Mitogen-activated protein kinase kinase inhibition enhances nuclear proapoptotic function of p53 in acute myelogenous leukemia cells.
- Authors
- Kojima K, Konopleva M, Samudio IJ, Ruvolo V, Andreeff M
- Journal
- Cancer Res - Page No 3210 (Sun Apr 01 00:00:00 EDT 2007)
- Volume (Year)
- 67 (2007)
- Product Usage
- -
Major differences in the responses of primary human leukocyte subsets to IFN-beta.
- Authors
- van Boxel-Dezaire AH, Zula JA, Xu Y, Ransohoff RM, Jacobberger JW, Stark GR
- Journal
- J Immunol - Page No 5888 (Mon Nov 15 00:00:00 EST 2010)
- Volume (Year)
- 185 (2010)
- Product Usage
- -
MDM2-dependent inhibition of p53 is required for Epstein-Barr virus B-cell growth transformation and infected-cell survival.
- Authors
- Forte E, Luftig MA
- Journal
- J Virol - Page No 2491 (Sun Mar 01 00:00:00 EST 2009)
- Volume (Year)
- 83 (2009)
- Product Usage
- -
Role of peroxisome proliferator-activated receptor-gamma and its coactivator DRIP205 in cellular responses to CDDO (RTA-401) in acute myelogenous leukemia.
- Authors
- Tsao T, Kornblau S, Safe S, Watt JC, Ruvolo V, Chen W, Qiu Y, Coombes KR, Ju Z, Abdelrahim M, Schober W, Ling X, Kardassis D, Meyer C, Schimmer A, Kantarjian H, Andreeff M, Konopleva M
- Journal
- Cancer Res - Page No 4949 (Tue Jun 15 00:00:00 EDT 2010)
- Volume (Year)
- 70 (2010)
- Product Usage
- -
Occurrence of urolithins, gut microbiota ellagic acid metabolites and proliferation markers expression response in the human prostate gland upon consumption of walnuts and pomegranate juice.
- Authors
- Gonzalez-Sarrias A, Gimenez-Bastida JA, Garcia-Conesa MT, Gomez-Sanchez MB, Garcia-Talavera NV, Gil-Izquierdo A, Sanchez-Alvarez C, Fontana-Compiano LO, Morga-Egea JP, Pastor-Quirante FA, Martinez-Diaz F, Tomas-Barberan FA, Espin JC
- Journal
- Mol Nutr Food Res - Page No 311 (Mon Mar 01 00:00:00 EST 2010)
- Volume (Year)
- 54 (2010)
- Product Usage
- -
Effects of model organophosphorous pesticides on DNA damage and proliferation of HepG2 cells.
- Authors
- Hreljac I, Zajc I, Lah T, Filipic M
- Journal
- Environ Mol Mutagen - Page No 360 (Sun Jun 01 00:00:00 EDT 2008)
- Volume (Year)
- 49 (2008)
- Product Usage
- -
Epidermal growth factor receptor and mutant p53 expand an esophageal cellular subpopulation capable of epithelial-to-mesenchymal transition through ZEB transcription factors.
- Authors
- Ohashi S, Natsuizaka M, Wong GS, Michaylira CZ, Grugan KD, Stairs DB, Kalabis J, Vega ME, Kalman RA, Nakagawa M, Klein-Szanto AJ, Herlyn M, Diehl JA, Rustgi AK, Nakagawa H
- Journal
- Cancer Res - Page No 4174 (Sat May 15 00:00:00 EDT 2010)
- Volume (Year)
- 70 (2010)
- Product Usage
- -
Involvement of the estrogen receptor beta in genistein-induced expression of p21(waf1/cip1) in PC-3 prostate cancer cells.
- Authors
- Matsumura K, Tanaka T, Kawashima H, Nakatani T
- Journal
- Anticancer Res - Page No 709 (Sat Mar 01 00:00:00 EST 2008)
- Volume (Year)
- 28 (2008)
- Product Usage
- -
The experimental chemotherapeutic N6-furfuryladenosine (kinetin-riboside) induces rapid ATP depletion, genotoxic stress, and CDKN1A(p21) upregulation in human cancer cell lines.
- Authors
- Cabello CM, Bair WB 3rd, Ley S, Lamore SD, Azimian S, Wondrak GT
- Journal
- Biochem Pharmacol - Page No 1125 (Wed Apr 01 00:00:00 EDT 2009)
- Volume (Year)
- 77 (2009)
- Product Usage
- -
Human Primary Articular Chondrocytes, Chondroblasts-Like Cells, and Dedifferentiated Chondrocytes: Differences in Gene, MicroRNA, and Protein Expression and Phenotype.
- Authors
- Karlsen TA, Shahdadfar A, Brinchmann JE
- Journal
- Tissue Eng Part C Methods - Page No (Thu Oct 21 00:00:00 EDT 2010)
- Volume (Year)
- (2010)
- Product Usage
- -
p21 gene polymorphisms in systemic lupus erythematosus.
- Authors
- Kong EK, Chong WP, Wong WH, Lau CS, Chan TM, Ng PK, Song YQ, Mak W, Lau YL
- Journal
- Rheumatology (Oxford) - Page No 220 (Thu Feb 01 00:00:00 EST 2007)
- Volume (Year)
- 46 (2007)
- Product Usage
- -
BEX2 regulates mitochondrial apoptosis and G1 cell cycle in breast cancer.
- Authors
- Naderi A, Liu J, Bennett IC
- Journal
- Int J Cancer - Page No 1596 (Thu Apr 01 00:00:00 EDT 2010)
- Volume (Year)
- 126 (2010)
- Product Usage
- -
Expression of an IFN-inducible cellular senescence gene, IFI16, is up-regulated by p53.
- Authors
- Song LL, Alimirah F, Panchanathan R, Xin H, Choubey D
- Journal
- Mol Cancer Res - Page No 1732 (Sat Nov 01 00:00:00 EDT 2008)
- Volume (Year)
- 6 (2008)
- Product Usage
- -
In vitro and in vivo cytotoxic effects of PRIMA-1 on hepatocellular carcinoma cells expressing mutant p53ser249.
- Authors
- Shi H, Lambert JM, Hautefeuille A, Bykov VJ, Wiman KG, Hainaut P, de Fromentel CC
- Journal
- Carcinogenesis - Page No 1428 (Tue Jul 01 00:00:00 EDT 2008)
- Volume (Year)
- 29 (2008)
- Product Usage
- -
Blockade of MGMT expression by O6 benzyl guanine leads to inhibition of pancreatic cancer growth and induction of apoptosis.
- Authors
- Konduri SD, Ticku J, Bobustuc GC, Sutphin RM, Colon J, Isley B, Bhakat KK, Srivenugopal KS, Baker CH
- Journal
- Clin Cancer Res - Page No 6087 (Thu Oct 01 00:00:00 EDT 2009)
- Volume (Year)
- 15 (2009)
- Product Usage
- -
Transformation of human mesenchymal stem cells in radiation carcinogenesis: long-term effect of ionizing radiation.
- Authors
- Christensen R, Alsner J, Brandt Sorensen F, Dagnaes-Hansen F, Kolvraa S, Serakinci N
- Journal
- Regen Med - Page No 849 (Sat Nov 01 00:00:00 EDT 2008)
- Volume (Year)
- 3 (2008)
- Product Usage
- -
Expression of CAR in SW480 and HepG2 cells during G1 is associated with cell proliferation.
- Authors
- Osabe M, Sugatani J, Takemura A, Yamazaki Y, Ikari A, Kitamura N, Negishi M, Miwa M
- Journal
- Biochem Biophys Res Commun - Page No 1027 (Fri May 16 00:00:00 EDT 2008)
- Volume (Year)
- 369 (2008)
- Product Usage
- -
Patterns of microcystin-LR induced alteration of the expression of genes involved in response to DNA damage and apoptosis.
- Authors
- Zegura B, Zajc I, Lah TT, Filipic M
- Journal
- Toxicon - Page No 615 (Sat Mar 15 00:00:00 EDT 2008)
- Volume (Year)
- 51 (2008)
- Product Usage
- -
High radiosensitivity of germ cells in human male fetus.
- Authors
- Lambrot R, Coffigny H, Pairault C, Lecureuil C, Frydman R, Habert R, Rouiller-Fabre V
- Journal
- J Clin Endocrinol Metab - Page No 2632 (Sun Jul 01 00:00:00 EDT 2007)
- Volume (Year)
- 92 (2007)
- Product Usage
- -
DNA damage, DNA repair rates and mRNA expression levels of cell cycle genes (TP53, p21CDKN1A, BCL2 and BAX) with respect to occupational exposure to styrene.
- Authors
- Hanova M, Vodickova L, Vaclavikova R, Smerhovsky Z, Stetina R, Hlavac P, Naccarati A, Slyskova J, Polakova V, Soucek P, Kumar R, Hemminki K, Vodicka P
- Journal
- Carcinogenesis - Page No (Thu Oct 21 00:00:00 EDT 2010)
- Volume (Year)
- (2010)
- Product Usage
- -
Anticancer mechanisms of doxorubicin and zoledronic acid in breast cancer tumor growth in bone.
- Authors
- Ottewell PD, Woodward JK, Lefley DV, Evans CA, Coleman RE, Holen I
- Journal
- Mol Cancer Ther - Page No 2821 (Thu Oct 01 00:00:00 EDT 2009)
- Volume (Year)
- 8 (2009)
- Product Usage
- -
MDM2 antagonists activate p53 and synergize with genotoxic drugs in B-cell chronic lymphocytic leukemia cells.
- Authors
- Coll-Mulet L, Iglesias-Serret D, Santidrian AF, Cosialls AM, de Frias M, Castano E, Campas C, Barragan M, de Sevilla AF, Domingo A, Vassilev LT, Pons G, Gil J
- Journal
- Blood - Page No 4109 (Mon May 15 00:00:00 EDT 2006)
- Volume (Year)
- 107 (2006)
- Product Usage
- -
Nucleotide excision repair gene expression after Cisplatin treatment in melanoma.
- Authors
- Bowden NA, Ashton KA, Avery-Kiejda KA, Zhang XD, Hersey P, Scott RJ
- Journal
- Cancer Res - Page No 7918 (Fri Oct 15 00:00:00 EDT 2010)
- Volume (Year)
- 70 (2010)
- Product Usage
- -
Impact of repeated bouts of eccentric exercise on myogenic gene expression.
- Authors
- Costa A, Dalloul H, Hegyesi H, Apor P, Csende Z, Racz L, Vaczi M, Tihanyi J
- Journal
- Eur J Appl Physiol - Page No 427 (Thu Nov 01 00:00:00 EDT 2007)
- Volume (Year)
- 101 (2007)
- Product Usage
- -
AHR- and DNA-damage-mediated gene expression responses induced by benzo(a)pyrene in human cell lines.
- Authors
- Hockley SL, Arlt VM, Brewer D, Te Poele R, Workman P, Giddings I, Phillips DH
- Journal
- Chem Res Toxicol - Page No 1797 (Sat Dec 01 00:00:00 EST 2007)
- Volume (Year)
- 20 (2007)
- Product Usage
- -
Ectopically hTERT expressing adult human mesenchymal stem cells are less radiosensitive than their telomerase negative counterpart.
- Authors
- Serakinci N, Christensen R, Graakjaer J, Cairney CJ, Keith WN, Alsner J, Saretzki G, Kolvraa S
- Journal
- Exp Cell Res - Page No 1056 (Sat Mar 10 00:00:00 EST 2007)
- Volume (Year)
- 313 (2007)
- Product Usage
- -
UVB radiation induces persistent activation of ribosome and oxidative phosphorylation pathways.
- Authors
- Tsai ML, Chang KY, Chiang CS, Shu WY, Weng TC, Chen CR, Huang CL, Lin HK, Hsu IC
- Journal
- Radiat Res - Page No 716 (Mon Jun 01 00:00:00 EDT 2009)
- Volume (Year)
- 171 (2009)
- Product Usage
- -
Effects of strength training with eccentric overload on muscle adaptation in male athletes.
- Authors
- Friedmann-Bette B, Bauer T, Kinscherf R, Vorwald S, Klute K, Bischoff D, Muller H, Weber MA, Metz J, Kauczor HU, Bartsch P, Billeter R
- Journal
- Eur J Appl Physiol - Page No 821 (Mon Mar 01 00:00:00 EST 2010)
- Volume (Year)
- 108 (2010)
- Product Usage
- -
5alpha,8alpha-Epidioxy-22E-ergosta-6,9(11),22-trien-3beta-ol from an edible mushroom suppresses growth of HL60 leukemia and HT29 colon adenocarcinoma cells.
- Authors
- Kobori M, Yoshida M, Ohnishi-Kameyama M, Takei T, Shinmoto H
- Journal
- Biol Pharm Bull - Page No 755 (Sat Apr 01 00:00:00 EST 2006)
- Volume (Year)
- 29 (2006)
- Product Usage
- -
Small bowel carcinoid (enterochromaffin cell) neoplasia exhibits transforming growth factor-beta1-mediated regulatory abnormalities including up-regulation of C-Myc and MTA1.
- Authors
- Kidd M, Modlin IM, Pfragner R, Eick GN, Champaneria MC, Chan AK, Camp RL, Mane SM
- Journal
- Cancer - Page No 2420 (Fri Jun 15 00:00:00 EDT 2007)
- Volume (Year)
- 109 (2007)
- Product Usage
- -
AKT and p21 WAF1/CIP1 as potential genistein targets in BRCA1-mutant human breast cancer cell lines.
- Authors
- Privat M, Aubel C, Arnould S, Communal Y, Ferrara M, Bignon YJ
- Journal
- Anticancer Res - Page No 2049 (Tue Jun 01 00:00:00 EDT 2010)
- Volume (Year)
- 30 (2010)
- Product Usage
- -
Cytogenetic instability in ovarian epithelial cells from women at risk of ovarian cancer.
- Authors
- Pejovic T, Yates JE, Liu HY, Hays LE, Akkari Y, Torimaru Y, Keeble W, Rathbun RK, Rodgers WH, Bale AE, Ameziane N, Zwaan CM, Errami A, Thuillier P, Cappuccini F, Olson SB, Cain JM, Bagby GC Jr
- Journal
- Cancer Res - Page No 9017 (Fri Sep 15 00:00:00 EDT 2006)
- Volume (Year)
- 66 (2006)
- Product Usage
- -
Modulatory effects of 1,25-dihydroxyvitamin D3 on human B cell differentiation.
- Authors
- Chen S, Sims GP, Chen XX, Gu YY, Chen S, Lipsky PE
- Journal
- J Immunol - Page No 1634 (Wed Aug 01 00:00:00 EDT 2007)
- Volume (Year)
- 179 (2007)
- Product Usage
- -
The up-regulation profiles of p21WAF1/CIP1 and RUNX1/AML1 correlate with myometrial infiltration in endometrioid endometrial carcinoma.
- Authors
- Planaguma J, Gonzalez M, Doll A, Monge M, Gil-Moreno A, Baro T, Garcia A, Xercavins J, Alameda F, Abal M, Reventos J
- Journal
- Hum Pathol - Page No 1050 (Tue Aug 01 00:00:00 EDT 2006)
- Volume (Year)
- 37 (2006)
- Product Usage
- -
Interactions of ErbB4 and Kap1 connect the growth factor and DNA damage response pathways.
- Authors
- Gilmore-Hebert M, Ramabhadran R, Stern DF
- Journal
- Mol Cancer Res - Page No 1388 (Fri Oct 01 00:00:00 EDT 2010)
- Volume (Year)
- 8 (2010)
- Product Usage
- -
Bone morphogenetic proteins and receptors are over-expressed in bone-marrow cells of multiple myeloma patients and support myeloma cells by inducing ID genes.
- Authors
- Grcevic D, Kusec R, Kovacic N, Lukic A, Lukic IK, Ivcevic S, Nemet D, Seiwerth RS, Ostojic SK, Croucher PI, Marusic A
- Journal
- Leuk Res - Page No 742 (Tue Jun 01 00:00:00 EDT 2010)
- Volume (Year)
- 34 (2010)
- Product Usage
- -
Skeletal muscle gene expression profiling in mitochondrial disorders.
- Authors
- Crimi M, Bordoni A, Menozzi G, Riva L, Fortunato F, Galbiati S, Del Bo R, Pozzoli U, Bresolin N, Comi GP
- Journal
- Faseb J - Page No 866 (Sun May 01 00:00:00 EDT 2005)
- Volume (Year)
- 19 (2005)
- Product Usage
- -
Phosphorylation of Thr18 and Ser20 of p53 in Ad-p53-induced apoptosis.
- Authors
- Nakamizo A, Amano T, Zhang W, Zhang XQ, Ramdas L, Liu TJ, Bekele BN, Shono T, Sasaki T, Benedict WF, Sawaya R, Lang FF
- Journal
- Neuro Oncol - Page No 275 (Sun Jun 01 00:00:00 EDT 2008)
- Volume (Year)
- 10 (2008)
- Product Usage
- -
Gene expression profiles modulated by the human carcinogen aristolochic acid I in human cancer cells and their dependence on TP53.
- Authors
- Simoes ML, Hockley SL, Schwerdtle T, da Costa GG, Schmeiser HH, Phillips DH, Arlt VM
- Journal
- Toxicol Appl Pharmacol - Page No 86 (Wed Oct 01 00:00:00 EDT 2008)
- Volume (Year)
- 232 (2008)
- Product Usage
- -
The cinnamon-derived Michael acceptor cinnamic aldehyde impairs melanoma cell proliferation, invasiveness, and tumor growth.
- Authors
- Cabello CM, Bair WB 3rd, Lamore SD, Ley S, Bause AS, Azimian S, Wondrak GT
- Journal
- Free Radic Biol Med - Page No 220 (Thu Jan 15 00:00:00 EST 2009)
- Volume (Year)
- 46 (2009)
- Product Usage
- -
c-myc antisense oligonucleotides sensitize human colorectal cancer cells to chemotherapeutic drugs.
- Authors
- Abaza MS, Al-Saffar A, Al-Sawan S, Al-Attiyah R
- Journal
- Tumour Biol - Page No 287 (Tue Jan 01 00:00:00 EST 2008)
- Volume (Year)
- 29 (2008)
- Product Usage
- -
Expression of DNA damage response genes indicate progressive breast tumors.
- Authors
- Gochhait S, Dar S, Pal R, Gupta P, Bamezai RN
- Journal
- Cancer Lett - Page No 305 (Sun Jan 18 00:00:00 EST 2009)
- Volume (Year)
- 273 (2009)
- Product Usage
- -
Cortactin modulates RhoA activation and expression of Cip/Kip cyclin-dependent kinase inhibitors to promote cell cycle progression in 11q13-amplified head and neck squamous cell carcinoma cells.
- Authors
- Croucher DR, Rickwood D, Tactacan CM, Musgrove EA, Daly RJ
- Journal
- Mol Cell Biol - Page No 5057 (Mon Nov 01 00:00:00 EDT 2010)
- Volume (Year)
- 30 (2010)
- Product Usage
- -
The anticancer agent prodigiosin induces p21WAF1/CIP1 expression via transforming growth factor-beta receptor pathway.
- Authors
- Soto-Cerrato V, Vinals F, Lambert JR, Perez-Tomas R
- Journal
- Biochem Pharmacol - Page No 1340 (Thu Nov 01 00:00:00 EDT 2007)
- Volume (Year)
- 74 (2007)
- Product Usage
- -
Hypoxia response and VEGF-A expression in human proximal tubular epithelial cells in stable and progressive renal disease.
- Authors
- Rudnicki M, Perco P, Enrich J, Eder S, Heininger D, Bernthaler A, Wiesinger M, Sarkozi R, Noppert SJ, Schramek H, Mayer B, Oberbauer R, Mayer G
- Journal
- Lab Invest - Page No 337 (Sun Mar 01 00:00:00 EST 2009)
- Volume (Year)
- 89 (2009)
- Product Usage
- -
Differential roles of STAT1alpha and STAT1beta in fludarabine-induced cell cycle arrest and apoptosis in human B cells.
- Authors
- Baran-Marszak F, Feuillard J, Najjar I, Le Clorennec C, Bechet JM, Dusanter-Fourt I, Bornkamm GW, Raphael M, Fagard R
- Journal
- Blood - Page No 2475 (Fri Oct 15 00:00:00 EDT 2004)
- Volume (Year)
- 104 (2004)
- Product Usage
- -