Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017882.hg.1 |
TC04001304.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017882.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017882.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011052.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011052.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011052.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011052.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011052.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011052.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011052.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011052.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011052.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011052.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011052.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011052.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011052.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011052.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011052.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017881.hg.1 |
TC04001304.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017881.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017881.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400175291.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400175291.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400175291.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400175291.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400175291.hg.1 |
TC0400011052.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400011052.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011052.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011052.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011052.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011052.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011052.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011052.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011052.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011052.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011052.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011052.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011052.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011052.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011052.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011052.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400175292.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400175292.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400175292.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400175292.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400175292.hg.1 |
TC0400011052.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400175293.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400175293.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400175293.hg.1 |
ENST00000264888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400175293.hg.1 |
NM_002416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400175293.hg.1 |
TC0400011052.hg.1 |
Clariom D TC |