A section of mouse intestine stained with a combination of fluorescent stains.
A section of mouse intestine stained with a combination of fluorescent stains. Fibronectin, an extracellular matrix adhesion molecule, was labeled using a chicken primary antibody against fibronectin and visualized using green-fluorescent Alexa Fluor® 488 Goat Anti–Chicken IgG antibody (Cat. No. A11039). The filamentous actin (F-actin) prevalent in the brush border was stained with red-fluorescent Alexa Fluor® 568 phalloidin (Cat. No. A12380). Finally, the nuclei were stained with DAPI (Cat. No. D1306, D3571, D21490).
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
Increase in accuracy and precision using the Qubit® Go ›
Performance of the Qubit® dsDNA HS Assay Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›