Simultaneous detection of smooth muscle actin and aquaporin 1 in a mouse mammary alveolus.
Simultaneous detection of smooth muscle actin and aquaporin 1 in a mouse mammary alveolus at day 1 of lactation. Smooth muscle actin of myoepithelial cells was identified with a mouse primary antibody and visualized with red-fluorescent Alexa Fluor® 594 Goat Anti–Mouse IgG antibody (Cat. No. A11005). Aquaporin 1 of endothelial cells was detected with a rabbit primary antibody and visualized with green-fluorescent Alexa Fluor® 488 Goat Anti–Rabbit IgG antibody (Cat. No. A11008). Image contributed by Jonathan Shillingford, Laboratory of Genetics and Physiology, National Institutes of Health, Bethesda, Maryland.
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›