Human iPSC staining using mouse anti beta-tubulin monoclonal antibody.
Human iPSCs were cultured on glass slides under feeder-free conditions in StemPro® hESC Medium (A1000701). Cells were fixed and permed with the Image-iT® Fixation/Permeabilization Kit (R37602). Oct4 (green) expression was visualized using anti-Oct4 primary Ab and Alexa Fluor® 488 secondary Ab (A11034). Tubulin (red) expression was visualized using anti-tubulin primary Ab (322600) and Alexa Fluor® 594 secondary Ab (A11005). Nuclei (blue) were labeled with NucBlue™ Fixed Cell Stain (R37606). Images were collected on the FLoid™ Cell Imaging Station (4471136).
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Phagocytosis in MMM Cells using pHrodo™ Red E. coli BioParticles® Conjugate® (Catalog # P35361) Go ›
Muntjac skin fibroblast labeled with rhodamine phalloidin. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
Increase in accuracy and precision using the Qubit® Go ›
Performance of the Qubit® dsDNA HS Assay Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›
Human iPSC staining. Go ›
Human iPSC staining using Mouse Anti Beta-Tubulin Monoclonal Antibody. Go ›