Catalog number: AMEX1000
Designed to eliminate the complexities of microscopy, the EVOS XL Core system captures high-quality brightfield cell images right at your benchtop within a minute. It’s the perfect instrument for monitoring cell cultures, either within a hood or in your cell culture room.
The EVOS XL Core system extends EVOS simplicity and ease of use to brightfield microscopy. With a bright LED light source, a high-resolution color camera, and a choice of objectives, it can capture, display, and save high-quality color images in just a few clicks.
The XL Core system offers:
Unpacking and installation in less than 2 minutes with no alignment or bulbs to change, ever
High-quality brightfield and phase contrast images with adjustable focus, light intensity, and color tone
Optional mechanical stage allows use of more objectives than the included fixed stage
Additional post-acquisition image analysis via the Thermo Fisher Connect™ cloud application
Many operational functions can be controlled through software, reducing the need to touch the instrument
Can be used within a biosafety cabinet or hood
Your choice whether to maximize speed or resolution
View more EVOS systems videos and demos under Resources.
Category | Attribute | Description |
Optics | Description | Infinity-corrected optical system; RMS-threaded objectives with 45 mm parfocal distance |
Illumination | Adjustable-intensity LED (>50,000-hour life per light cube) | |
Contrast methods | Transmitted light (bright field and phase contrast) | |
Objectives | 4x, 10x, 20x, 40x, plan LWD, infinity; Optional 2x (no phase) | |
Objective turret | 4-position, manual control | |
Condenser | 3-position turret for bright field and phase contrast |
|
Condenser working distance | 60 mm | |
Stages | Fixed stage (AMEX1000); Optional mechanical stage (AMEX4712) • X-Y axis controls and vessel holder framework • 120 mm x 78 mm range of motion • Both knobs 56 mm/rev |
|
Focus mechanism | Coaxial focus knobs with tension control: • Coarse focus: 38 mm/rev • Fine focus: 0.2 mm/rev, precision 0.002 mm |
|
LCD Display | 12.1” high-resolution (1,024 x 768 pixels) color monitor with adjustable tilt | |
Camera | Color: 1/2” 2,048 x 1, 536, 3.1 megapixels | |
Physical characteristics | Power supply | AC adapter; input 100-240 V, 47-63 Hz; output 12 V DC/2.0 A 24 W max |
Instrument dimensions |
21.0” (W) x 6.0” (H) x 12.5” (D) 533 mm (W) x 406 mm (H) x 318 mm (D) |
|
Weight | 9.1 kg (20.1 lb) 10.0 kg (22.1 lb) with mechanical stage |
|
Output ports | 2 USB 2.0 ports |
|
Hardware/Software | Image acquisition | Embedded operating system with imaging software for image capture and save via mouse or front-mounted manual buttons |
Captured images | Color camera: 24-bit full-color TIFF or PNG; JPEG, BMP (2,048 x 1,536 pixels) |
For Research Use Only. Not for use in diagnostic procedures.