Debbie Bowe
Sr. Tech Specialist and Master Trainer
Environmental and Process Monitoring
Meet our techs: Debbie Bowe
Debbie enjoys helping customers solve problems in all areas including programming samplers, passing leak checks and getting flow audits to meet standards.
Our techs are here to answer your questions, just email us.
If you like this month's Q&A, check out our Talk Tech archive.
Question
Answer
Recent changes to Microsoft Windows Security interfere with the installer program. You must disable 'Windows firewall'. If you have a wireless network adapter, you must disable the wireless adapter temporarily.
For more information, see TB09132013, Windows 7 and Windows 8 Software Compatibility Technical Bulletin.
Don't miss out. Subscribe today.
Talk Tech is published here every month. Receive our monthly Talk Tech answers delivered right to your inbox by subscribing using the button below.