Advanced battery materials analysis
In battery production and research, the quality of materials is becoming critical. Small contaminants in the NCM powder, for example, can have disastrous results in the final product. To trace these contaminants effectively, high-resolution SEM imaging with EDS analysis for chemistry is needed. When fully automated, this combination is a powerful tool for powder quality inspection.
Each particle class can be labeled with a conductance of the particles, allowing you to sort on conductance of particles. This allows you to assess the impact of contamination much more accurately since a small organic contamination is not as severe as a metallic conductive contamination.
To view overall chemistry of the particle population, a ternary diagram can be generated where all particles are represented. With Ni, Co, and Mn on each axis, the outyears and general trends can be seen instantly.
ChemiSEM Technology
Thermo Scientific ChemiSEM Technology revolutionizes and simplifies EDS analysis by fully integrating SEM and EDS functions into a single, cohesive user interface. Based on live quantification and building on decades of expertise in EDS analysis, the technology provides elemental information quickly and easily, guaranteeing reliable results in less time. ChemiSEM Technology now comes with a powerful new feature: ChemiPhase. ChemiPhase identifies unique phases with a big data approach, finding minor and trace elements while eliminating user bias and reducing possible mistakes.
Electron optical |
|
Electron optical magnification range |
|
Light optical magnification |
|
Resolution |
|
Image resolution options |
|
Acceleration voltages |
|
Vacuum levels |
|
Detector |
|
Sample size |
|
Sample loading time |
|
Desktop SEM in battery development
See how the Phenom ParticleX Battery Desktop SEM performs in the real world with application notes, webinars, and more.
Lithium Ion Battery Analysis blog post
SEM Particle Analysis for Battery Manufacturing webinar
Desktop SEM Blogs
Want to unlock the great power of scanning electron microscopy without having to compromise on usability? Enhance your knowledge on Scanning Electron Microscopy and find out how Desktop SEM can optimally support your research in our Phenom Desktop SEM blogs.
Desktop scanning electron microscopes for industrial manufacturing
The product selector is here to assist you in choosing the most suitable Scanning Electron Microscope (SEM) system and software for your research. Find out in a few minutes which SEM best suits your research application:
Watch on-demand: Particle Analysis Applications Using Desktop SEM Webinar Series
In each on-demand webinar, our expert will focus on one particular analysis application and how the Phenom ParticleX Desktop SEM overcomes some of the most common challenges. See the abstracts for individual sessions below.
- How to certify your NCM powder quality with SEM+EDS
- Electron-microscope-grade cleanliness in electronics
- Speed up your automated gunshot residue analysis
- Technical cleanliness analysis with an SEM: Why?
- Understand your steel with automated inclusion analysis
- One tool for complete AM powder characterization
On-demand webinar: New Phenom ParticleX Steel Desktop SEM
Watch our on-demand webinar to learn more about:
- Industry-specific software and automation designed specifically for inclusion analysis in metallurgic samples
- Small footprint, which allows it be added to any analytical lab without the need to adjust facility infrastructure
- Easy-to-use interface and sample loading that allow you to begin using the instrument with minimal training
- High-quality, in-house SEM quality control
On-demand webinar: New Phenom ParticleX Steel Desktop SEM
Watch our on-demand webinar to learn more about:
- Industry-specific software and automation designed specifically for inclusion analysis in metallurgic samples
- Small footprint, which allows it be added to any analytical lab without the need to adjust facility infrastructure
- Easy-to-use interface and sample loading that allow you to begin using the instrument with minimal training
- High-quality, in-house SEM quality control
Watch Phenom ParticleX Desktop SEM: Quality Starts at the Microscale
Watch this on-demand webinar to learn how Phenom ParticleX desktop SEMs are the ideal solution for:
- ISO 16232 and VDA-19 Compliance for Technical Cleanliness.
- ISO 4406 and ISO 4407 for contamination of hydraulic fluids.
- Particle morphology for additive manufacturing feedstocks.
- Workflow examples for particle classifications and reporting.
Webinar: Scanning electron microscopy: selecting the right technology for your needs
This on-demand webinar has been designed to help you decide which SEM best meets your unique needs. We present an overview of Thermo Fisher Scientific SEM technology for multi-user research labs and focus on how these wide-ranging solutions deliver performance, versatility, in situ dynamics and faster time to results. Watch this webinar if you are interested in:
- How the needs for different microanalysis modalities are met (EDX, EBSD, WDS, CL, etc.).
- How samples are characterized in their natural state without the need for sample preparation.
- How new advanced automation allows researchers to save time and increase productivity.
ParticleX Steel Desktop SEM - Workflow introduction
Watch on-demand: Particle Analysis Applications Using Desktop SEM Webinar Series
In each on-demand webinar, our expert will focus on one particular analysis application and how the Phenom ParticleX Desktop SEM overcomes some of the most common challenges. See the abstracts for individual sessions below.
- How to certify your NCM powder quality with SEM+EDS
- Electron-microscope-grade cleanliness in electronics
- Speed up your automated gunshot residue analysis
- Technical cleanliness analysis with an SEM: Why?
- Understand your steel with automated inclusion analysis
- One tool for complete AM powder characterization
On-demand webinar: New Phenom ParticleX Steel Desktop SEM
Watch our on-demand webinar to learn more about:
- Industry-specific software and automation designed specifically for inclusion analysis in metallurgic samples
- Small footprint, which allows it be added to any analytical lab without the need to adjust facility infrastructure
- Easy-to-use interface and sample loading that allow you to begin using the instrument with minimal training
- High-quality, in-house SEM quality control
On-demand webinar: New Phenom ParticleX Steel Desktop SEM
Watch our on-demand webinar to learn more about:
- Industry-specific software and automation designed specifically for inclusion analysis in metallurgic samples
- Small footprint, which allows it be added to any analytical lab without the need to adjust facility infrastructure
- Easy-to-use interface and sample loading that allow you to begin using the instrument with minimal training
- High-quality, in-house SEM quality control
Watch Phenom ParticleX Desktop SEM: Quality Starts at the Microscale
Watch this on-demand webinar to learn how Phenom ParticleX desktop SEMs are the ideal solution for:
- ISO 16232 and VDA-19 Compliance for Technical Cleanliness.
- ISO 4406 and ISO 4407 for contamination of hydraulic fluids.
- Particle morphology for additive manufacturing feedstocks.
- Workflow examples for particle classifications and reporting.
Webinar: Scanning electron microscopy: selecting the right technology for your needs
This on-demand webinar has been designed to help you decide which SEM best meets your unique needs. We present an overview of Thermo Fisher Scientific SEM technology for multi-user research labs and focus on how these wide-ranging solutions deliver performance, versatility, in situ dynamics and faster time to results. Watch this webinar if you are interested in:
- How the needs for different microanalysis modalities are met (EDX, EBSD, WDS, CL, etc.).
- How samples are characterized in their natural state without the need for sample preparation.
- How new advanced automation allows researchers to save time and increase productivity.
ParticleX Steel Desktop SEM - Workflow introduction
Process control using electron microscopy
Modern industry demands high throughput with superior quality, a balance that is maintained through robust process control. SEM and TEM tools with dedicated automation software provide rapid, multi-scale information for process monitoring and improvement.
Quality control and failure analysis
Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
Fundamental Materials Research
Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
Technical Cleanliness
More than ever, modern manufacturing necessitates reliable, quality components. With scanning electron microscopy, parts cleanliness analysis can be brought inhouse, providing you with a broad range of analytical data and shortening your production cycle.
EDS Elemental Analysis
Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
3D EDS Tomography
Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
Atomic-Scale Elemental Mapping with EDS
Atomic-resolution EDS provides unparalleled chemical context for materials analysis by differentiating the elemental identity of individual atoms. When combined with high-resolution TEM, it is possible to observe the precise organization of atoms in a sample.
Particle analysis
Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
EDS Elemental Analysis
Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
3D EDS Tomography
Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
Atomic-Scale Elemental Mapping with EDS
Atomic-resolution EDS provides unparalleled chemical context for materials analysis by differentiating the elemental identity of individual atoms. When combined with high-resolution TEM, it is possible to observe the precise organization of atoms in a sample.
Particle analysis
Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.