Real-time PCR products with high analytical sensitivity are needed to support laboratory research to study outbreaks and determine disease etiology where pathogens are similar.
Applied Biosystems real-time PCR gastrointestinal research products are used to analyze a range of GI pathogens including viruses, bacteria, and parasites, with high analytical sensitivity. These solutions are designed to provide efficient, informative results and help solve many of the challenges associated with the use of immunoassay-based, culture-based, and other molecular research methods.
Applied Biosystems TrueMark real-time PCR solutions are part of a sample-to-results workflow. Benefits include the ability to:
- Detect and differentiate multiple pathogens
- Obtain reliable real-time PCR results—Accurate, sensitive, and specific
- Select from multiple format options to suit your testing capacity—Choose from single tube assays, 96- or 384-well plates, cards, or OpenArrayplates
- Order individual assays, predesigned panels, or customize your own—Select from a catalogue of >600 assays targeting pathogens or resistance genes
- Reduce time-to-implementation—Accelerate and streamline your validation process with Analytical Validation Consulting Services
TrueMark Enteric Pathogen Select Panels
- Single-tube multiplexed PCR assay for simultaneous detection of multiple enteric pathogens
- Enables superior analytical sensitivity and faster time-to-results than traditional methods like culture and microscopy
- Enables research laboratories to test for genetic traces of multiple enteric pathogens in one stool sample
- Each panel comes with dedicated amplification control and recommended process control and sample prep
Bacterial Panel I |
Shigella / EIEC |
Salmonella |
Campylobacter |
STEC |
Bacterial Panel II |
Yersinia |
Vibrio |
Aeromonas |
Plesiomonas shigelloides |
Viral Panel |
Adenovirus F 40/41 |
Astrovirus |
Rotavirus A |
Norovirus Panel |
Norovirus GI |
Norovirus GII |
Parasitic Panel |
Cryptosporidium |
E. histolytica |
Giardia lamblia |
Cyclospora |
TrueMark Enteric Pathogen Expanded Panel—TaqMan Array Card format | ||
Number of wells | 384 | |
Number of targets | 22 enteric plus 2 controls (Xeno and Bacilus atrophaeus) | |
Number of samples | 8 | |
Master mix volume | 400 μL per array card (384 reactions) | |
Reaction volume | 1 μL | |
Samples per day | Up to 64 | |
Compatible instruments | QuantStudio 7, 7 Pro, and 12K Flex Real-Time PCR systems with array card block | |
Time-to-results | 4 hours |
TrueMark Enteric Pathogen Expanded Panel—TaqMan OpenArray format | ||
Number of wells | 2,668 | |
Number of targets | 26 enteric plus 2 controls (Xeno and Bacillus atrophaeus) | |
Number of samples | Up to 48 samples per plate (spotted in duplicate) | |
Master mix volume | 120 µL per OpenArray plate | |
Reaction volume | 33 nL | |
Samples per day | ||
Compatible instruments | QuantStudio 12K Flex Real-Time PCR System with OpenArray block | |
Time-to-results | 5 hours |
Don't see what you're looking for?
Whether you want to modify from an existing design or start from scratch, we're here to walk you through the process. We offer customization across all formats to help you create a tailored solution that meets your laboratory's throughput and business needs.
Ordering information
Product |
|
TrueMark Enteric Pathogen Expanded Panel – OpenArray format | |
TrueMark Enteric Pathogen Expanded Panel –TAC format | |
Custom OpenArray plates | |
TrueMark Infectious Disease Custom Gastrointestinal Panel | |
TrueMark Custom Single tube assays for Enteric Pathogen Detection | |
TrueMark Enteric Pathogen Select Panels |
For Research Use Only. Not for use in diagnostic procedures.
Complete your infectious disease workflow with a single kit for DNA and RNA extraction
Maximize your nucleic acid yield for greater assay sensitivity from a range of biological specimens for infectious disease applications with the MagMAX Prime Viral/Pathogen NA Isolation Kit.