Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602060 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMPRSS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMPRSS2 - transmembrane protease, serine 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135099.1 | 612 | Missense Mutation | ATG,GTG | M,V 197 | NP_001128571.1 | |
NM_005656.3 | 612 | Missense Mutation | ATG,GTG | M,V 160 | NP_005647.3 | |
XM_005261043.2 | 612 | Missense Mutation | ATG,GTG | M,V 120 | XP_005261100.1 | |
XM_011529731.2 | 612 | Missense Mutation | ATG,GTG | M,V 160 | XP_011528033.1 | |
XM_011529732.2 | 612 | Missense Mutation | ATG,GTG | M,V 160 | XP_011528034.1 | |
XM_011529733.1 | 612 | Missense Mutation | ATG,GTG | M,V 52 | XP_011528035.1 |
More Information
Set Membership: |
HapMap |