TSV Manual Defrost Freezers
TSV Manual Defrost Freezers

TSV Manual Defrost Freezers

The new Thermo Scientific™ TSV 18 cu ft Manual Defrost Laboratory Freezer offers an economical and environmentally friendly solution for safe and reliable general-purpose storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
TSV18FPSAGeneral purpose
TSV05FPSAGeneral Purpose Undercounter Freezer
Catalog number TSV18FPSA
Price (USD)/ Each
-
Request A Quote
Type:
General purpose
The Thermo Scientific™ TSV Value manual defrost laboratory freezers offer an economical, simple, and environmentally friendly solution for safe and reliable general-purpose storage.
  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally friendly, SNAP compliant hydrocarbon refrigerants and foam
  • Adjustable temperature control
  • Easy to clearn, durable finish
Specifications
TypeGeneral purpose
Capacity (English)18 cu. ft.
Temperature Range-10°C to -22°C
Temperature SensorNTC Probe
Temperature (Setpoint)-20°C
Door StyleSolid Swing
No. of Doors1
Cabinet MaterialPainted Steel
Chamber MaterialCold-rolled Steel
InsulationCyclopentane (C5H10)
InteriorABS
Exterior FinishEnameled steel
Shelves6
Certifications/ComplianceProp 65
StyleUpright
DisplayDigital
CastersNo casters. Only 2 rollers at rear and 2 front adjustable legs
ColorWhite
Condenser TypeForced air cooled
For Use With (Application)Non-Corrosive, Non-Flammable, Non-Explosive, Good Air Ventilation
PortsNo
Warranty13 months
DefrostManual
LidNone
Min. ClearanceUnit Exterior Dimensions (W x D x H) 777 x 705 x 1895 mm (30.6 x 27.8 x 74.6 in.) Installation Clearance of 12 in. at the top, 2 in. at the rear and 2 in. on each side.
ModelTSV
Monitoring OptionsExternal
Setpoint SecurityNo
Amperage5.5A
Plug TypeNEMA 5-15 P
Voltage115 V
Phase1 Phase
Energy Usage4 kw-hr/day
CompressorHermetic
RefrigerantR290, 110g / Flammable
HoldsRacks
Rack Capacity10
Unit SizeEach