TMCS Silylation Reagent
TMCS Silylation Reagent
Thermo Scientific™
TMCS Silylation Reagent
Catalyze the derivatization of difficult-to-silylate compounds using Thermo Scientific™ TMCS Silylation Reagent.
Have Questions?
Catalog number TS-88530
Price (USD)/ Each
386.82
-
Add to cart
For Use With:
GC
Price (USD)/ Each
386.82
Add to cart
TMCS Silylation Reagent
Catalog numberTS-88530
Price (USD)/ Each
386.82
-
Add to cart
Catalyze the derivatization of difficult-to-silylate compounds using Thermo Scientific™ TMCS Silylation Reagent. It is an excellent adjunct for forming TMS ethers for GC determinations. TMCS is also useful for preparing TMS derivatives of organic acids.
  • Excellent adjunct for forming trimethylsilyl ethers for GC determinations
  • Used to prepare TMS derivatives of organic acids
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use WithGC
Standard Concentration98 to 100%
HazardsFlammable, Toxic
StorageRoom Temperature
Quantity25 g
Standard ComponentsChlorotrimethylsilane
Unit SizeEach