Find additional information here: Thermo Scientific™ Talos™ Arctica™ is a 200kV FEG transmission and scanning electron microscope (S/TEM)
The Thermo Scientific™ Talos™ Arctica™ is a 200kV FEG transmission and scanning electron microscope (S/TEM). It is a powerful, stable, and versatile system for delivering high-resolution 3D characterization of biological and biomaterials samples in cell biology, structural biology, and nanotechnology research. The Talos S/TEM enables scientists to quickly obtain better insight and understanding of macromolecular structures, cellular components, cells, and tissues in three dimensions.
Fastest 200 kV FEG S/TEM for sample analysis in multiple dimensions
The Talos TEM is a new generation in TEM, built to deliver rapid access to 2D and 3D data so that you can concentrate on discovery. The system's constant-power, C-Twin lens delivers outstanding optical performance to ensure an optimal balance of contrast and resolution. The Talos TEM's ultra-stable platform includes a piezo stage with stable electronics and a rugged system enclosure for maximum thermal and mechanical stability.
Talos Arctica TEM Benefits for Structural Biology
Increased data acquisition speed
Acquisition speeds have been improved up to 3 times for tomography and up to 1.5 times for single particle analysis when EPU is used compared to side entry loading systems.
High data throughput
Robotic sample handling and automated loading for up to 12 samples enhances up to 40% in data throughput.
Unattended operation
Autofilling of liquid nitrogen and powerful applications SW allows for continuous unattended platform operation and automated data acquisition.
Low cost of ownership
New design allows for remote diagnostics and preventive service, lowering ownership cost and maximizing systems uptime.