Tubulin Tracker™ Deep Red
Tubulin Tracker™ Deep Red
Invitrogen™

Tubulin Tracker™ Deep Red

Tubulin Tracker Deep Red provides deep-red/far-red fluorescence when bound to polymerized tubulin in live cells. Tubulin Tracker Deep Red isRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
T3407760 slides
T34076300 slides
Catalog number T34077
Price (USD)/ 60 slides
181.00
-
Add to cart
Quantity:
60 slides
Price (USD)/ 60 slides
181.00
Add to cart
Tubulin Tracker™ Deep Red
Catalog numberT34077
Price (USD)/ 60 slides
181.00
-
Add to cart
Tubulin Tracker Deep Red provides deep-red/far-red fluorescence when bound to polymerized tubulin in live cells. Tubulin Tracker Deep Red is based on Docetaxel conjugated with a bright, photostable deep-red fluorophore. Docetaxel belongs to the family of cytoskeletal drugs that target tubulin. Tubulin Tracker Deep Red absorbs and emits optimally at 652 nm and 669 nm, respectively and can be visualized with standard Cy5 filter settings using almost any fluorescent imaging instrument. It can be multiplexed with blue, green, orange, red, and near-IR fluorophores.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Excitation Wavelength Range652⁄669
Quantity60 slides
Product LineTubulinTracker™
Shipping ConditionApproved for shipment on Wet or Dry Ice
Label TypeUV-Vis Labels
Product TypeTubulin Tracker
SubCellular LocalizationTubulin
Unit Size60 slides
Contents & Storage
Contains sufficient material for at least 60 slides at 1 mL per slide.

•1 vial Tubulin Tracker Deep Red (Add 60 μL/vial of anhydrous DMSO to make a 1000X (1mM) stock solution.  Dissolve stock solution 1:1000 in cell imaging solution to make 1X (1 μM) staining solution.)
•1 x 77 mg Probenecid (Dissolve vial content in 1 mL of live cell compatible buffer to make a 100X stock solution.)

Store at ≤–20°C after recieving.
Desiccate and protect from light.
When stored as directed, product is stable for at least 6 months.