CSPD™ Substrate (0.4 mM Ready-To-Use) with Emerald-II™ Enhancer, Size A
CSPD™ Substrate (0.4 mM Ready-To-Use) with Emerald-II™ Enhancer, Size A
Invitrogen™
CSPD™ Substrate (0.4 mM Ready-To-Use) with Emerald-II™ Enhancer, Size A
This CSPD™ substrate with Emerald-II™ Enhancer is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting onRead more
Have Questions?
Catalog number T2212
Price (USD)/ 100 ml
794.00
-
Add to cart
Request bulk or custom format
Price (USD)/ 100 ml
794.00
Add to cart
CSPD™ Substrate (0.4 mM Ready-To-Use) with Emerald-II™ Enhancer, Size A
Catalog numberT2212
Price (USD)/ 100 ml
794.00
-
Add to cart
This CSPD™ substrate with Emerald-II™ Enhancer is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.
This CSPD™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.
• Chemiluminescent substrates provide highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
• Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.
This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours. Solution assays require chemiluminescent enhancers. Membrane-based assays may also require them for increased light output and sensitivity.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeELISA Substrate
Detection MethodChemiluminescence
FormLiquid
FormatBottle
Product LineCSPD™, NovaBright™
Substrate TypeAlkaline Phosphatase
Shipping ConditionRoom Temperature
Unit Size100 ml
Contents & Storage
Store the product at 4°C.