Specifications
Chemical Name or MaterialEthanol
pH7 @ 20°C 10 g/l aq.sol
Melting Point-114°C
Absorbance<=1.00 @ 205 NM, <=0.20 @ 220 NM, <=0.10 @ 230 NM, <=0.015 @ 254 NM
ColorColorless
AppearanceClear, Colorless Liquid
Boiling Point78°C
Quantity1 L
CAS64-17-5
PackagingSafe-Cote™ Amber Glass
Residue after Evaporation<=1 ppm
Molecular FormulaC2 H6 O
Titratable Acid<=0.0005 mEq/g
Titratable Base<=0.0002 mEq/g
Water<=0.2%
SynonymEthyl alcohol; Absolute ethanol
Molecular Weight (g/mol)46.07
Percent Purity≥99.5%
GradeMolecular Biology
Unit SizeCase of 6